Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 16478779-16479032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM134B
Entrez Gene ID
54463
Gene Name
family with sequence similarity 134 member B
Gene Aliases
JK-1, JK1
UniGene
Hs.481704
Gene Chromosome Location
Chr.5: 16473038-16617058 on Build GRCh38
Transcripts
NM_001034850.2 NM_019000.4 XM_011514053.2 XM_011514054.2 XM_011514055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3248177 c.385G>T p.E129* Chr.5: 16478850-16478850 on Build GRCh38
6488335 c.302T>A p.I101N Chr.5: 16478933-16478933 on Build GRCh38
345729 c.674T>G p.L225R Chr.5: 16478984-16478984 on Build GRCh38
6393255 c.317A>G p.K106R Chr.5: 16478918-16478918 on Build GRCh38
6587918 c.364C>T p.Q122* Chr.5: 16478871-16478871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373543417 Chr.5: 16478822-16478822 on Build GRCh38
rs767329180 Chr.5: 16478908-16478908 on Build GRCh38
rs761925091 Chr.5: 16478854-16478854 on Build GRCh38
rs748217475 Chr.5: 16478971-16478971 on Build GRCh38
rs766150181 Chr.5: 16479001-16479001 on Build GRCh38
rs367886987 Chr.5: 16478948-16478948 on Build GRCh38
rs748279839 Chr.5: 16478840-16478840 on Build GRCh38
rs368052487 Chr.5: 16478844-16478844 on Build GRCh38
rs370570919 Chr.5: 16478812-16478812 on Build GRCh38
rs750191157 Chr.5: 16478971-16478972 on Build GRCh38
rs141215735 Chr.5: 16479003-16479003 on Build GRCh38
rs761822553 Chr.5: 16478835-16478835 on Build GRCh38
rs772338562 Chr.5: 16478821-16478821 on Build GRCh38
rs773070026 Chr.5: 16478997-16478997 on Build GRCh38
rs531943235 Chr.5: 16478965-16478965 on Build GRCh38
rs374340868 Chr.5: 16478861-16478861 on Build GRCh38
rs754610295 Chr.5: 16478896-16478896 on Build GRCh38
rs200587445 Chr.5: 16478853-16478853 on Build GRCh38
rs779603632 Chr.5: 16478811-16478811 on Build GRCh38
rs768725177 Chr.5: 16478959-16478959 on Build GRCh38
rs375825002 Chr.5: 16478926-16478926 on Build GRCh38
rs368759467 Chr.5: 16478862-16478862 on Build GRCh38
rs370636928 Chr.5: 16478855-16478855 on Build GRCh38
rs747446867 Chr.5: 16478955-16478955 on Build GRCh38
rs1021020925 Chr.5: 16478834-16478834 on Build GRCh38
rs751185980 Chr.5: 16478883-16478883 on Build GRCh38
rs200070690 Chr.5: 16478947-16478947 on Build GRCh38
rs184749223 Chr.5: 16478839-16478839 on Build GRCh38
rs950385242 Chr.5: 16478891-16478891 on Build GRCh38
rs372401503 Chr.5: 16478913-16478913 on Build GRCh38
rs564789943 Chr.5: 16478870-16478870 on Build GRCh38
rs752223454 Chr.5: 16478911-16478911 on Build GRCh38
rs947997843 Chr.5: 16478996-16478996 on Build GRCh38
rs991974394 Chr.5: 16478945-16478945 on Build GRCh38
rs769879385 Chr.5: 16478985-16478985 on Build GRCh38
rs765165944 Chr.5: 16478947-16478949 on Build GRCh38
rs370743879 Chr.5: 16478823-16478823 on Build GRCh38
rs762831720 Chr.5: 16479000-16479000 on Build GRCh38
rs746380834 Chr.5: 16478935-16478935 on Build GRCh38

View Full Product Details