Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22055540-22055783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMAS
Entrez Gene ID
55907
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Gene Aliases
CSS
UniGene
Hs.311346
Gene Chromosome Location
Chr.12: 22046174-22065674 on Build GRCh38
Transcripts
NM_018686.5 NR_135117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1511433 c.524G>A p.R175H Chr.12: 22055575-22055575 on Build GRCh38
6071730 c.559+1G>T p.? Chr.12: 22055611-22055611 on Build GRCh38
1299237 c.503C>G p.S168C Chr.12: 22055554-22055554 on Build GRCh38
1742465 c.512C>T p.S171F Chr.12: 22055563-22055563 on Build GRCh38
2005548 c.493G>T p.G165* Chr.12: 22055544-22055544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199957467 Chr.12: 22055575-22055575 on Build GRCh38
rs968937528 Chr.12: 22055576-22055576 on Build GRCh38
rs770899077 Chr.12: 22055642-22055642 on Build GRCh38
rs888735618 Chr.12: 22055738-22055738 on Build GRCh38
rs371050975 Chr.12: 22055574-22055574 on Build GRCh38
rs978646578 Chr.12: 22055586-22055586 on Build GRCh38
rs139792913 Chr.12: 22055668-22055668 on Build GRCh38
rs941519758 Chr.12: 22055743-22055743 on Build GRCh38
rs759425087 Chr.12: 22055657-22055657 on Build GRCh38
rs760231041 Chr.12: 22055629-22055629 on Build GRCh38
rs187337997 Chr.12: 22055687-22055687 on Build GRCh38
rs546600015 Chr.12: 22055698-22055709 on Build GRCh38
rs776531882 Chr.12: 22055648-22055648 on Build GRCh38
rs77214419 Chr.12: 22055677-22055677 on Build GRCh38
rs368101382 Chr.12: 22055624-22055624 on Build GRCh38
rs771685299 Chr.12: 22055614-22055614 on Build GRCh38
rs1047327411 Chr.12: 22055688-22055688 on Build GRCh38
rs976841479 Chr.12: 22055672-22055672 on Build GRCh38
rs747784089 Chr.12: 22055609-22055609 on Build GRCh38
rs966533556 Chr.12: 22055595-22055595 on Build GRCh38
rs534806772 Chr.12: 22055697-22055697 on Build GRCh38

View Full Product Details