Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19573068-19573321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf62
Entrez Gene ID
57020
Gene Name
chromosome 16 open reading frame 62
Gene Aliases
-
UniGene
Hs.654964
Gene Chromosome Location
Chr.16: 19555415-19701163 on Build GRCh38
Transcripts
NM_001300743.1 NM_020314.5 XM_005255435.3 XM_006721065.3 XM_011545896.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4643953 c.652A>G p.T218A Chr.16: 19573218-19573218 on Build GRCh38
967919 c.640C>A p.L214I Chr.16: 19573206-19573206 on Build GRCh38
305554 c.324C>T p.S108S Chr.16: 19573157-19573157 on Build GRCh38
967917 c.628C>T p.R210W Chr.16: 19573194-19573194 on Build GRCh38
5747714 c.649T>C p.Y217H Chr.16: 19573215-19573215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763894705 Chr.16: 19573159-19573159 on Build GRCh38
rs752063213 Chr.16: 19573273-19573273 on Build GRCh38
rs143649089 Chr.16: 19573125-19573125 on Build GRCh38
rs377476012 Chr.16: 19573259-19573259 on Build GRCh38
rs534959158 Chr.16: 19573138-19573138 on Build GRCh38
rs147199562 Chr.16: 19573158-19573158 on Build GRCh38
rs756324639 Chr.16: 19573234-19573240 on Build GRCh38
rs144556424 Chr.16: 19573227-19573227 on Build GRCh38
rs926517177 Chr.16: 19573150-19573150 on Build GRCh38
rs762440421 Chr.16: 19573265-19573265 on Build GRCh38
rs761403020 Chr.16: 19573251-19573251 on Build GRCh38
rs370715531 Chr.16: 19573168-19573168 on Build GRCh38
rs201379369 Chr.16: 19573250-19573250 on Build GRCh38
rs746865470 Chr.16: 19573226-19573226 on Build GRCh38
rs186236295 Chr.16: 19573160-19573160 on Build GRCh38
rs1024843921 Chr.16: 19573205-19573205 on Build GRCh38
rs539532149 Chr.16: 19573264-19573264 on Build GRCh38
rs570834372 Chr.16: 19573137-19573137 on Build GRCh38
rs1050421133 Chr.16: 19573277-19573277 on Build GRCh38
rs758948904 Chr.16: 19573177-19573177 on Build GRCh38
rs28688709 Chr.16: 19573293-19573293 on Build GRCh38
rs34128516 Chr.16: 19573235-19573235 on Build GRCh38
rs536088496 Chr.16: 19573217-19573217 on Build GRCh38
rs375466958 Chr.16: 19573206-19573206 on Build GRCh38
rs373505689 Chr.16: 19573188-19573188 on Build GRCh38
rs781381888 Chr.16: 19573108-19573108 on Build GRCh38
rs989162977 Chr.16: 19573229-19573229 on Build GRCh38
rs982554310 Chr.16: 19573278-19573278 on Build GRCh38
rs932947259 Chr.16: 19573236-19573236 on Build GRCh38
rs1008986629 Chr.16: 19573213-19573213 on Build GRCh38
rs780519985 Chr.16: 19573184-19573184 on Build GRCh38
rs775464952 Chr.16: 19573157-19573157 on Build GRCh38
rs139739334 Chr.16: 19573123-19573123 on Build GRCh38
rs749909431 Chr.16: 19573170-19573170 on Build GRCh38
rs140487282 Chr.16: 19573194-19573194 on Build GRCh38
rs775554738 Chr.16: 19573218-19573218 on Build GRCh38
rs756307949 Chr.16: 19573198-19573198 on Build GRCh38
rs748328502 Chr.16: 19573195-19573195 on Build GRCh38
rs766942376 Chr.16: 19573269-19573269 on Build GRCh38
rs751981247 Chr.16: 19573186-19573186 on Build GRCh38
rs768918170 Chr.16: 19573100-19573100 on Build GRCh38
rs772184499 Chr.16: 19573216-19573216 on Build GRCh38
rs751355959 Chr.16: 19573208-19573214 on Build GRCh38
rs755417874 Chr.16: 19573283-19573283 on Build GRCh38
rs199767367 Chr.16: 19573212-19573212 on Build GRCh38
rs144895626 Chr.16: 19573169-19573169 on Build GRCh38
rs149026004 Chr.16: 19573257-19573257 on Build GRCh38

View Full Product Details