Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55544245-55544472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP36
Entrez Gene ID
112942
Gene Name
cilia and flagella associated protein 36
Gene Aliases
BARTL1, CCDC104
UniGene
Hs.264208
Gene Chromosome Location
Chr.2: 55519595-55545080 on Build GRCh38
Transcripts
NM_001282761.1 NM_080667.6 XM_011532499.1 XM_017003254.1 XM_017003255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094910 c.810G>A p.R270R Chr.2: 55544252-55544252 on Build GRCh38
3053378 c.894A>C p.E298D Chr.2: 55544261-55544261 on Build GRCh38
6496325 c.955C>T p.Q319* Chr.2: 55544322-55544322 on Build GRCh38
1021886 c.808C>T p.R270W Chr.2: 55544250-55544250 on Build GRCh38
5058970 c.902T>C p.L301P Chr.2: 55544269-55544269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751084654 Chr.2: 55544420-55544420 on Build GRCh38
rs764667361 Chr.2: 55544284-55544284 on Build GRCh38
rs374786938 Chr.2: 55544359-55544359 on Build GRCh38
rs755086122 Chr.2: 55544331-55544331 on Build GRCh38
rs778241181 Chr.2: 55544352-55544352 on Build GRCh38
rs374822899 Chr.2: 55544390-55544390 on Build GRCh38
rs779350603 Chr.2: 55544332-55544332 on Build GRCh38
rs767240767 Chr.2: 55544305-55544305 on Build GRCh38
rs748634021 Chr.2: 55544337-55544337 on Build GRCh38
rs762497638 Chr.2: 55544285-55544285 on Build GRCh38
rs866393195 Chr.2: 55544302-55544302 on Build GRCh38
rs374899371 Chr.2: 55544274-55544274 on Build GRCh38
rs754242168 Chr.2: 55544299-55544299 on Build GRCh38
rs369833243 Chr.2: 55544406-55544406 on Build GRCh38
rs756282373 Chr.2: 55544322-55544322 on Build GRCh38
rs1010798593 Chr.2: 55544381-55544381 on Build GRCh38
rs144603204 Chr.2: 55544317-55544317 on Build GRCh38
rs766581168 Chr.2: 55544397-55544397 on Build GRCh38
rs1128735 Chr.2: 55544355-55544355 on Build GRCh38
rs754833608 Chr.2: 55544404-55544405 on Build GRCh38
rs748816756 Chr.2: 55544374-55544374 on Build GRCh38
rs905035268 Chr.2: 55544394-55544394 on Build GRCh38
rs765552870 Chr.2: 55544326-55544327 on Build GRCh38
rs372141624 Chr.2: 55544358-55544358 on Build GRCh38
rs921449888 Chr.2: 55544283-55544283 on Build GRCh38
rs1004184343 Chr.2: 55544272-55544272 on Build GRCh38
rs779779814 Chr.2: 55544354-55544354 on Build GRCh38
rs200593801 Chr.2: 55544328-55544328 on Build GRCh38
rs138513616 Chr.2: 55544304-55544304 on Build GRCh38
rs754082200 Chr.2: 55544402-55544402 on Build GRCh38
rs780991734 Chr.2: 55544406-55544408 on Build GRCh38
rs751509022 Chr.2: 55544298-55544298 on Build GRCh38
rs753172304 Chr.2: 55544418-55544418 on Build GRCh38
rs768199043 Chr.2: 55544379-55544379 on Build GRCh38
rs934286079 Chr.2: 55544361-55544361 on Build GRCh38
rs769457156 Chr.2: 55544278-55544278 on Build GRCh38
rs774167318 Chr.2: 55544385-55544385 on Build GRCh38
rs756394401 Chr.2: 55544282-55544282 on Build GRCh38
rs992618915 Chr.2: 55544307-55544307 on Build GRCh38
rs750608095 Chr.2: 55544342-55544358 on Build GRCh38
rs138348471 Chr.2: 55544326-55544326 on Build GRCh38
rs765298042 Chr.2: 55544408-55544408 on Build GRCh38
rs760431071 Chr.2: 55544392-55544392 on Build GRCh38
rs763978612 Chr.2: 55544291-55544291 on Build GRCh38
rs745535635 Chr.2: 55544353-55544353 on Build GRCh38

View Full Product Details