Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62605994-62606253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML3
Entrez Gene ID
256364
Gene Name
echinoderm microtubule associated protein like 3
Gene Aliases
ELP95
UniGene
Hs.379785
Gene Chromosome Location
Chr.11: 62602218-62613020 on Build GRCh38
Transcripts
NM_001300793.1 NM_001300794.1 NM_153265.2 XM_005273878.3 XM_006718489.3 XM_006718490.3 XM_006718491.3 XM_011544896.2 XM_011544897.2 XM_017017480.1 XM_017017481.1 XM_017017482.1 XM_017017483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653819 c.1600C>T p.R534W Chr.11: 62606119-62606119 on Build GRCh38
4871613 c.1607G>A p.R536H Chr.11: 62606112-62606112 on Build GRCh38
5498667 c.1540G>A p.E514K Chr.11: 62606179-62606179 on Build GRCh38
4824425 c.1603G>C p.D535H Chr.11: 62606116-62606116 on Build GRCh38
3451018 c.1606C>T p.R536C Chr.11: 62606113-62606113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866158226 Chr.11: 62606093-62606093 on Build GRCh38
rs762775845 Chr.11: 62606152-62606152 on Build GRCh38
rs767569389 Chr.11: 62606165-62606165 on Build GRCh38
rs142385289 Chr.11: 62606024-62606024 on Build GRCh38
rs770437837 Chr.11: 62606136-62606136 on Build GRCh38
rs968299265 Chr.11: 62606140-62606140 on Build GRCh38
rs776245964 Chr.11: 62606045-62606045 on Build GRCh38
rs768820507 Chr.11: 62606156-62606156 on Build GRCh38
rs759488783 Chr.11: 62606053-62606053 on Build GRCh38
rs774254864 Chr.11: 62606162-62606162 on Build GRCh38
rs777119904 Chr.11: 62606123-62606123 on Build GRCh38
rs767947780 Chr.11: 62606086-62606086 on Build GRCh38
rs746710321 Chr.11: 62606134-62606134 on Build GRCh38
rs750671374 Chr.11: 62606173-62606173 on Build GRCh38
rs757388631 Chr.11: 62606079-62606079 on Build GRCh38
rs745593458 Chr.11: 62606143-62606143 on Build GRCh38
rs372585430 Chr.11: 62606039-62606039 on Build GRCh38
rs1040906376 Chr.11: 62606031-62606031 on Build GRCh38
rs751058570 Chr.11: 62606050-62606051 on Build GRCh38
rs996361127 Chr.11: 62606107-62606107 on Build GRCh38
rs756232112 Chr.11: 62606122-62606122 on Build GRCh38
rs150541468 Chr.11: 62606186-62606186 on Build GRCh38
rs376428089 Chr.11: 62606020-62606020 on Build GRCh38
rs751856890 Chr.11: 62606077-62606077 on Build GRCh38
rs749802659 Chr.11: 62606103-62606103 on Build GRCh38
rs775549093 Chr.11: 62606056-62606056 on Build GRCh38
rs764066798 Chr.11: 62606068-62606068 on Build GRCh38
rs757978135 Chr.11: 62606104-62606104 on Build GRCh38
rs754438929 Chr.11: 62606185-62606185 on Build GRCh38
rs140559608 Chr.11: 62606182-62606182 on Build GRCh38
rs756695880 Chr.11: 62606091-62606091 on Build GRCh38
rs575047153 Chr.11: 62606144-62606144 on Build GRCh38
rs761172762 Chr.11: 62606114-62606138 on Build GRCh38
rs375226751 Chr.11: 62606231-62606231 on Build GRCh38
rs146863965 Chr.11: 62606100-62606100 on Build GRCh38
rs780477268 Chr.11: 62606129-62606129 on Build GRCh38
rs762109104 Chr.11: 62606164-62606164 on Build GRCh38
rs761180605 Chr.11: 62606174-62606174 on Build GRCh38
rs765096886 Chr.11: 62606055-62606055 on Build GRCh38
rs746621718 Chr.11: 62606030-62606030 on Build GRCh38
rs780975436 Chr.11: 62606137-62606137 on Build GRCh38
rs202159023 Chr.11: 62606151-62606151 on Build GRCh38
rs377089335 Chr.11: 62606090-62606090 on Build GRCh38
rs763086125 Chr.11: 62606060-62606060 on Build GRCh38

View Full Product Details