Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223002517-223002790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISP1
Entrez Gene ID
84976
Gene Name
dispatched RND transporter family member 1
Gene Aliases
DISPA
UniGene
Hs.528817
Gene Chromosome Location
Chr.1: 222814514-223005995 on Build GRCh38
Transcripts
NM_032890.3 XM_005273335.2 XM_006711592.2 XM_006711594.3 XM_011510072.2 XM_011510073.2 XM_011510074.2 XM_011510075.2 XM_011510077.2 XM_017002611.1 XM_017002612.1 XM_017002613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
341842 c.1197G>T p.W399C Chr.1: 223002594-223002594 on Build GRCh38
4526247 c.1383G>A p.G461G Chr.1: 223002780-223002780 on Build GRCh38
6440268 c.1303G>A p.D435N Chr.1: 223002700-223002700 on Build GRCh38
1339101 c.1306T>C p.F436L Chr.1: 223002703-223002703 on Build GRCh38
1960113 c.1188A>G p.P396P Chr.1: 223002585-223002585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538321379 Chr.1: 223002595-223002595 on Build GRCh38
rs764941613 Chr.1: 223002720-223002720 on Build GRCh38
rs949010743 Chr.1: 223002573-223002573 on Build GRCh38
rs766466751 Chr.1: 223002618-223002618 on Build GRCh38
rs756593395 Chr.1: 223002642-223002642 on Build GRCh38
rs374481235 Chr.1: 223002729-223002729 on Build GRCh38
rs776731383 Chr.1: 223002715-223002715 on Build GRCh38
rs774576194 Chr.1: 223002668-223002668 on Build GRCh38
rs760687407 Chr.1: 223002605-223002605 on Build GRCh38
rs143019748 Chr.1: 223002756-223002756 on Build GRCh38
rs370192171 Chr.1: 223002702-223002702 on Build GRCh38
rs200646799 Chr.1: 223002585-223002585 on Build GRCh38
rs754759315 Chr.1: 223002623-223002623 on Build GRCh38
rs757971360 Chr.1: 223002728-223002728 on Build GRCh38
rs370525290 Chr.1: 223002753-223002753 on Build GRCh38
rs773414429 Chr.1: 223002693-223002693 on Build GRCh38
rs753652728 Chr.1: 223002622-223002622 on Build GRCh38
rs190037827 Chr.1: 223002734-223002734 on Build GRCh38
rs978886358 Chr.1: 223002581-223002581 on Build GRCh38
rs757853636 Chr.1: 223002635-223002635 on Build GRCh38
rs371163462 Chr.1: 223002544-223002544 on Build GRCh38
rs768929196 Chr.1: 223002662-223002662 on Build GRCh38
rs752301395 Chr.1: 223002634-223002634 on Build GRCh38
rs749629171 Chr.1: 223002644-223002644 on Build GRCh38
rs140829703 Chr.1: 223002575-223002575 on Build GRCh38
rs777257995 Chr.1: 223002640-223002640 on Build GRCh38
rs767054825 Chr.1: 223002643-223002643 on Build GRCh38
rs146869710 Chr.1: 223002551-223002551 on Build GRCh38
rs138859731 Chr.1: 223002727-223002727 on Build GRCh38
rs567212527 Chr.1: 223002545-223002545 on Build GRCh38
rs772491387 Chr.1: 223002684-223002684 on Build GRCh38
rs770349473 Chr.1: 223002557-223002557 on Build GRCh38
rs781617125 Chr.1: 223002549-223002549 on Build GRCh38
rs143819396 Chr.1: 223002626-223002626 on Build GRCh38
rs746577969 Chr.1: 223002641-223002641 on Build GRCh38
rs114523965 Chr.1: 223002719-223002719 on Build GRCh38
rs916140567 Chr.1: 223002552-223002552 on Build GRCh38
rs575391513 Chr.1: 223002675-223002675 on Build GRCh38
rs773217795 Chr.1: 223002604-223002604 on Build GRCh38
rs774472223 Chr.1: 223002579-223002579 on Build GRCh38
rs775816983 Chr.1: 223002567-223002567 on Build GRCh38
rs749704141 Chr.1: 223002574-223002574 on Build GRCh38
rs763180722 Chr.1: 223002739-223002739 on Build GRCh38
rs760875536 Chr.1: 223002701-223002701 on Build GRCh38

View Full Product Details