Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59618721-59618994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT3
Entrez Gene ID
9245
Gene Name
glucosaminyl (N-acetyl) transferase 3, mucin type
Gene Aliases
C2/4GnT, C24GNT, C2GNT2, C2GNTM, GNTM
UniGene
Hs.194710
Gene Chromosome Location
Chr.15: 59611783-59620011 on Build GRCh38
Transcripts
NM_004751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
963472 c.636C>T p.D212D Chr.15: 59618874-59618874 on Build GRCh38
3502819 c.520G>A p.E174K Chr.15: 59618758-59618758 on Build GRCh38
6001663 c.587T>C p.V196A Chr.15: 59618825-59618825 on Build GRCh38
963473 c.705C>T p.S235S Chr.15: 59618943-59618943 on Build GRCh38
4505531 c.695C>T p.P232L Chr.15: 59618933-59618933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762904598 Chr.15: 59618952-59618952 on Build GRCh38
rs746647237 Chr.15: 59618818-59618818 on Build GRCh38
rs776828333 Chr.15: 59618938-59618938 on Build GRCh38
rs573657149 Chr.15: 59618790-59618790 on Build GRCh38
rs146888471 Chr.15: 59618784-59618784 on Build GRCh38
rs896399026 Chr.15: 59618747-59618747 on Build GRCh38
rs968838731 Chr.15: 59618961-59618961 on Build GRCh38
rs750345935 Chr.15: 59618860-59618860 on Build GRCh38
rs537114592 Chr.15: 59618764-59618764 on Build GRCh38
rs766247416 Chr.15: 59618956-59618956 on Build GRCh38
rs760912677 Chr.15: 59618850-59618850 on Build GRCh38
rs989440515 Chr.15: 59618917-59618917 on Build GRCh38
rs146608277 Chr.15: 59618870-59618870 on Build GRCh38
rs144448812 Chr.15: 59618864-59618864 on Build GRCh38
rs780910767 Chr.15: 59618965-59618965 on Build GRCh38
rs754607070 Chr.15: 59618883-59618883 on Build GRCh38
rs753574676 Chr.15: 59618852-59618852 on Build GRCh38
rs141256510 Chr.15: 59618792-59618792 on Build GRCh38
rs199814262 Chr.15: 59618924-59618924 on Build GRCh38
rs200969029 Chr.15: 59618858-59618858 on Build GRCh38
rs368185469 Chr.15: 59618967-59618967 on Build GRCh38
rs768801647 Chr.15: 59618928-59618928 on Build GRCh38
rs747878682 Chr.15: 59618891-59618891 on Build GRCh38
rs757048580 Chr.15: 59618857-59618857 on Build GRCh38
rs150778745 Chr.15: 59618904-59618904 on Build GRCh38
rs771411777 Chr.15: 59618894-59618894 on Build GRCh38
rs141022876 Chr.15: 59618888-59618888 on Build GRCh38
rs751059524 Chr.15: 59618882-59618882 on Build GRCh38
rs148722358 Chr.15: 59618763-59618763 on Build GRCh38
rs190663869 Chr.15: 59618958-59618958 on Build GRCh38
rs771501479 Chr.15: 59618832-59618832 on Build GRCh38
rs775997022 Chr.15: 59618861-59618861 on Build GRCh38
rs767616296 Chr.15: 59618841-59618841 on Build GRCh38
rs34785094 Chr.15: 59618800-59618800 on Build GRCh38
rs749690262 Chr.15: 59618825-59618825 on Build GRCh38
rs200140304 Chr.15: 59618895-59618895 on Build GRCh38
rs764994654 Chr.15: 59618946-59618946 on Build GRCh38
rs569867643 Chr.15: 59618762-59618762 on Build GRCh38
rs775472787 Chr.15: 59618845-59618845 on Build GRCh38
rs745309174 Chr.15: 59618806-59618806 on Build GRCh38
rs767563281 Chr.15: 59618768-59618768 on Build GRCh38
rs760023511 Chr.15: 59618834-59618834 on Build GRCh38
rs75826312 Chr.15: 59618780-59618780 on Build GRCh38
rs555465014 Chr.15: 59618773-59618773 on Build GRCh38
rs763499549 Chr.15: 59618877-59618877 on Build GRCh38
rs778834381 Chr.15: 59618801-59618801 on Build GRCh38
rs954222365 Chr.15: 59618885-59618885 on Build GRCh38
rs750254057 Chr.15: 59618951-59618951 on Build GRCh38
rs757920928 Chr.15: 59618814-59618814 on Build GRCh38
rs779487918 Chr.15: 59618817-59618817 on Build GRCh38
rs753665618 Chr.15: 59618794-59618794 on Build GRCh38
rs762080217 Chr.15: 59618940-59618940 on Build GRCh38
rs758660668 Chr.15: 59618825-59618825 on Build GRCh38
rs371193925 Chr.15: 59618925-59618925 on Build GRCh38
rs140710881 Chr.15: 59618819-59618819 on Build GRCh38

View Full Product Details