Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 376527-376798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHRR
Entrez Gene ID
57491
Gene Name
aryl-hydrocarbon receptor repressor
Gene Aliases
AHH, AHHR, bHLHe77
UniGene
Hs.50823
Gene Chromosome Location
Chr.5: 304176-438291 on Build GRCh38
Transcripts
NM_001242412.1 NM_020731.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
737858 c.258C>A p.V86V Chr.5: 376611-376611 on Build GRCh38
5868483 c.352C>A p.L118M Chr.5: 376705-376705 on Build GRCh38
4999413 c.341T>C p.L114P Chr.5: 376694-376694 on Build GRCh38
1067845 c.288G>A p.A96A Chr.5: 376641-376641 on Build GRCh38
6597067 c.295G>A p.A99T Chr.5: 376648-376648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051844136 Chr.5: 376735-376735 on Build GRCh38
rs752865107 Chr.5: 376630-376630 on Build GRCh38
rs749664194 Chr.5: 376574-376574 on Build GRCh38
rs1030472563 Chr.5: 376677-376677 on Build GRCh38
rs191789057 Chr.5: 376645-376645 on Build GRCh38
rs184336270 Chr.5: 376660-376660 on Build GRCh38
rs386684252 Chr.5: 376553-376554 on Build GRCh38
rs564461726 Chr.5: 376757-376757 on Build GRCh38
rs72711353 Chr.5: 376554-376554 on Build GRCh38
rs761568250 Chr.5: 376609-376609 on Build GRCh38
rs757564795 Chr.5: 376641-376641 on Build GRCh38
rs202003342 Chr.5: 376690-376690 on Build GRCh38
rs750823042 Chr.5: 376689-376689 on Build GRCh38
rs771583884 Chr.5: 376586-376586 on Build GRCh38
rs902731372 Chr.5: 376597-376597 on Build GRCh38
rs373559034 Chr.5: 376659-376659 on Build GRCh38
rs776606556 Chr.5: 376604-376604 on Build GRCh38
rs75015688 Chr.5: 376553-376553 on Build GRCh38
rs572863304 Chr.5: 376611-376611 on Build GRCh38
rs200873629 Chr.5: 376614-376614 on Build GRCh38
rs199848058 Chr.5: 376656-376656 on Build GRCh38
rs746976255 Chr.5: 376595-376595 on Build GRCh38
rs770847467 Chr.5: 376762-376762 on Build GRCh38
rs757630557 Chr.5: 376578-376578 on Build GRCh38
rs551220430 Chr.5: 376588-376588 on Build GRCh38
rs751205967 Chr.5: 376721-376721 on Build GRCh38
rs534361598 Chr.5: 376648-376648 on Build GRCh38
rs753283882 Chr.5: 376559-376559 on Build GRCh38
rs918515104 Chr.5: 376657-376657 on Build GRCh38
rs778312532 Chr.5: 376571-376571 on Build GRCh38
rs528568058 Chr.5: 376759-376759 on Build GRCh38
rs201485569 Chr.5: 376734-376734 on Build GRCh38
rs540294382 Chr.5: 376763-376763 on Build GRCh38
rs929795985 Chr.5: 376566-376566 on Build GRCh38
rs780707454 Chr.5: 376728-376728 on Build GRCh38
rs1000181466 Chr.5: 376769-376769 on Build GRCh38
rs545588638 Chr.5: 376741-376741 on Build GRCh38
rs919343460 Chr.5: 376747-376747 on Build GRCh38
rs369339217 Chr.5: 376632-376632 on Build GRCh38
rs376227888 Chr.5: 376647-376647 on Build GRCh38
rs765620811 Chr.5: 376680-376680 on Build GRCh38
rs529565718 Chr.5: 376580-376580 on Build GRCh38
rs762814318 Chr.5: 376658-376658 on Build GRCh38
rs115805955 Chr.5: 376771-376771 on Build GRCh38
rs201403478 Chr.5: 376612-376612 on Build GRCh38
rs769814891 Chr.5: 376740-376740 on Build GRCh38
rs537154972 Chr.5: 376631-376631 on Build GRCh38
rs745500296 Chr.5: 376731-376733 on Build GRCh38
rs368685976 Chr.5: 376557-376558 on Build GRCh38
rs745455415 Chr.5: 376582-376582 on Build GRCh38
rs146357233 Chr.5: 376770-376770 on Build GRCh38
rs367921340 Chr.5: 376644-376644 on Build GRCh38
rs201010505 Chr.5: 376675-376675 on Build GRCh38
rs768196917 Chr.5: 376653-376653 on Build GRCh38
rs754462794 Chr.5: 376725-376725 on Build GRCh38
rs748063530 Chr.5: 376655-376655 on Build GRCh38
rs754366584 Chr.5: 376640-376640 on Build GRCh38
rs1013474183 Chr.5: 376638-376638 on Build GRCh38
rs188027980 Chr.5: 376772-376772 on Build GRCh38
rs35008248 Chr.5: 376694-376694 on Build GRCh38
rs767740278 Chr.5: 376616-376616 on Build GRCh38
rs768512175 Chr.5: 376596-376596 on Build GRCh38
rs866630885 Chr.5: 376688-376688 on Build GRCh38

View Full Product Details