Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43375360-43375589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD4
Entrez Gene ID
53828
Gene Name
FXYD domain containing ion transport regulator 4
Gene Aliases
CHIF
UniGene
Hs.130497
Gene Chromosome Location
Chr.10: 43371644-43376335 on Build GRCh38
Transcripts
NM_001184963.1 NM_173160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4932642 c.130T>G p.C44G Chr.10: 43375531-43375531 on Build GRCh38
918163 c.121G>A p.G41R Chr.10: 43375522-43375522 on Build GRCh38
1658942 c.133G>A p.G45R Chr.10: 43375534-43375534 on Build GRCh38
4909978 c.146C>A p.A49D Chr.10: 43375547-43375547 on Build GRCh38
1745801 c.171G>A p.L57L Chr.10: 43375572-43375572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990863320 Chr.10: 43375409-43375409 on Build GRCh38
rs372862247 Chr.10: 43375561-43375561 on Build GRCh38
rs756495558 Chr.10: 43375475-43375475 on Build GRCh38
rs145486243 Chr.10: 43375562-43375562 on Build GRCh38
rs565869091 Chr.10: 43375557-43375557 on Build GRCh38
rs972754156 Chr.10: 43375468-43375468 on Build GRCh38
rs931550317 Chr.10: 43375482-43375482 on Build GRCh38
rs1004774125 Chr.10: 43375443-43375443 on Build GRCh38
rs762848271 Chr.10: 43375480-43375480 on Build GRCh38
rs773190513 Chr.10: 43375476-43375476 on Build GRCh38
rs750709866 Chr.10: 43375541-43375541 on Build GRCh38
rs769968604 Chr.10: 43375563-43375563 on Build GRCh38
rs146768172 Chr.10: 43375502-43375502 on Build GRCh38
rs997666876 Chr.10: 43375385-43375385 on Build GRCh38
rs754662151 Chr.10: 43375560-43375560 on Build GRCh38
rs754433020 Chr.10: 43375526-43375526 on Build GRCh38
rs767571808 Chr.10: 43375516-43375516 on Build GRCh38
rs779353769 Chr.10: 43375539-43375539 on Build GRCh38
rs780784925 Chr.10: 43375457-43375457 on Build GRCh38
rs548369294 Chr.10: 43375431-43375431 on Build GRCh38
rs769734332 Chr.10: 43375475-43375475 on Build GRCh38
rs11598898 Chr.10: 43375390-43375390 on Build GRCh38
rs963189667 Chr.10: 43375553-43375553 on Build GRCh38
rs369585516 Chr.10: 43375472-43375472 on Build GRCh38
rs1030027531 Chr.10: 43375399-43375399 on Build GRCh38
rs764268489 Chr.10: 43375523-43375523 on Build GRCh38
rs146396887 Chr.10: 43375563-43375563 on Build GRCh38
rs768140608 Chr.10: 43375451-43375451 on Build GRCh38
rs140339086 Chr.10: 43375534-43375534 on Build GRCh38
rs773968830 Chr.10: 43375490-43375490 on Build GRCh38
rs116641013 Chr.10: 43375565-43375565 on Build GRCh38
rs187142408 Chr.10: 43375521-43375521 on Build GRCh38
rs200599002 Chr.10: 43375522-43375522 on Build GRCh38
rs958542566 Chr.10: 43375402-43375402 on Build GRCh38
rs770764286 Chr.10: 43375486-43375486 on Build GRCh38
rs747545393 Chr.10: 43375464-43375464 on Build GRCh38
rs746720555 Chr.10: 43375449-43375449 on Build GRCh38

View Full Product Details