Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48693761-48694020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT1
Entrez Gene ID
904
Gene Name
cyclin T1
Gene Aliases
CCNT, CYCT1, HIVE1
UniGene
Hs.92308
Gene Chromosome Location
Chr.12: 48688458-48716998 on Build GRCh38
Transcripts
NM_001240.3 NM_001277842.1 XM_017020197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6136875 c.1406G>A p.G469E Chr.12: 48693808-48693808 on Build GRCh38
3968234 c.1432G>T p.E478* Chr.12: 48693782-48693782 on Build GRCh38
1676923 c.1453A>C p.K485Q Chr.12: 48693761-48693761 on Build GRCh38
3727190 c.1276C>T p.L426F Chr.12: 48693938-48693938 on Build GRCh38
6623099 c.1437G>A p.E479E Chr.12: 48693777-48693777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762789240 Chr.12: 48693932-48693932 on Build GRCh38
rs780374398 Chr.12: 48693792-48693792 on Build GRCh38
rs1053011061 Chr.12: 48693923-48693923 on Build GRCh38
rs748625270 Chr.12: 48693858-48693858 on Build GRCh38
rs754145543 Chr.12: 48693847-48693847 on Build GRCh38
rs756261087 Chr.12: 48693810-48693810 on Build GRCh38
rs201044557 Chr.12: 48693962-48693962 on Build GRCh38
rs763989714 Chr.12: 48693936-48693936 on Build GRCh38
rs775289744 Chr.12: 48693921-48693921 on Build GRCh38
rs61755289 Chr.12: 48693795-48693795 on Build GRCh38
rs3013 Chr.12: 48693873-48693873 on Build GRCh38
rs925606156 Chr.12: 48693859-48693859 on Build GRCh38
rs776385227 Chr.12: 48693804-48693804 on Build GRCh38
rs765093934 Chr.12: 48693805-48693805 on Build GRCh38
rs751567115 Chr.12: 48693938-48693938 on Build GRCh38
rs755236447 Chr.12: 48693852-48693852 on Build GRCh38
rs750482345 Chr.12: 48693807-48693807 on Build GRCh38
rs149726428 Chr.12: 48693833-48693833 on Build GRCh38
rs759231919 Chr.12: 48693893-48693893 on Build GRCh38
rs772728567 Chr.12: 48693860-48693860 on Build GRCh38
rs778472029 Chr.12: 48693868-48693868 on Build GRCh38
rs769436332 Chr.12: 48693898-48693898 on Build GRCh38
rs201423333 Chr.12: 48693796-48693796 on Build GRCh38
rs752564300 Chr.12: 48693806-48693806 on Build GRCh38
rs772879712 Chr.12: 48693800-48693800 on Build GRCh38
rs755256066 Chr.12: 48693967-48693967 on Build GRCh38
rs748982249 Chr.12: 48693837-48693837 on Build GRCh38
rs776091944 Chr.12: 48693875-48693875 on Build GRCh38
rs777279868 Chr.12: 48693999-48693999 on Build GRCh38
rs771638484 Chr.12: 48693799-48693799 on Build GRCh38
rs780131624 Chr.12: 48693827-48693827 on Build GRCh38
rs891688299 Chr.12: 48693925-48693925 on Build GRCh38
rs998118621 Chr.12: 48693945-48693945 on Build GRCh38
rs779337228 Chr.12: 48693855-48693855 on Build GRCh38
rs561483800 Chr.12: 48693963-48693963 on Build GRCh38
rs866465000 Chr.12: 48693886-48693886 on Build GRCh38
rs376740356 Chr.12: 48693801-48693801 on Build GRCh38

View Full Product Details