Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116856293-116856548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
NM_001242699.1 XM_005269815.3 XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866978 c.301G>T p.V101L Chr.10: 116856495-116856495 on Build GRCh38
6518323 c.353C>A p.P118H Chr.10: 116856547-116856547 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs917176737 Chr.10: 116856523-116856523 on Build GRCh38
rs968914490 Chr.10: 116856463-116856463 on Build GRCh38
rs890697582 Chr.10: 116856416-116856416 on Build GRCh38
rs574389793 Chr.10: 116856440-116856440 on Build GRCh38
rs760203244 Chr.10: 116856502-116856502 on Build GRCh38
rs897906418 Chr.10: 116856412-116856412 on Build GRCh38
rs767310157 Chr.10: 116856448-116856448 on Build GRCh38
rs993634943 Chr.10: 116856434-116856434 on Build GRCh38
rs189065497 Chr.10: 116856494-116856494 on Build GRCh38
rs150427030 Chr.10: 116856470-116856470 on Build GRCh38
rs955844641 Chr.10: 116856499-116856499 on Build GRCh38
rs140471319 Chr.10: 116856464-116856464 on Build GRCh38
rs1040172679 Chr.10: 116856338-116856338 on Build GRCh38
rs547842948 Chr.10: 116856443-116856443 on Build GRCh38
rs185397190 Chr.10: 116856368-116856368 on Build GRCh38
rs1030969735 Chr.10: 116856386-116856386 on Build GRCh38
rs1031473934 Chr.10: 116856482-116856482 on Build GRCh38
rs897954983 Chr.10: 116856328-116856328 on Build GRCh38
rs766190886 Chr.10: 116856519-116856519 on Build GRCh38
rs1037911374 Chr.10: 116856326-116856326 on Build GRCh38
rs992723309 Chr.10: 116856515-116856515 on Build GRCh38
rs1007668526 Chr.10: 116856432-116856432 on Build GRCh38
rs993756355 Chr.10: 116856377-116856377 on Build GRCh38

View Full Product Details