Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1557643-1557880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP8
Entrez Gene ID
1850
Gene Name
dual specificity phosphatase 8
Gene Aliases
C11orf81, HB5, HVH-5, HVH8
UniGene
Hs.41688
Gene Chromosome Location
Chr.11: 1554051-1572839 on Build GRCh38
Transcripts
NM_004420.2 XM_011519932.2 XM_011519933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7119622 c.750C>G p.I250M Chr.11: 1557865-1557865 on Build GRCh38
5913174 c.742G>A p.A248T Chr.11: 1557873-1557873 on Build GRCh38
6888838 c.792C>T p.T264T Chr.11: 1557823-1557823 on Build GRCh38
7183265 c.743C>G p.A248G Chr.11: 1557872-1557872 on Build GRCh38
6650409 c.768C>T p.I256I Chr.11: 1557847-1557847 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534452918 Chr.11: 1557696-1557696 on Build GRCh38
rs775298923 Chr.11: 1557764-1557764 on Build GRCh38
rs779190227 Chr.11: 1557805-1557805 on Build GRCh38
rs753157729 Chr.11: 1557783-1557783 on Build GRCh38
rs113161544 Chr.11: 1557788-1557788 on Build GRCh38
rs781269302 Chr.11: 1557808-1557808 on Build GRCh38
rs774055480 Chr.11: 1557834-1557834 on Build GRCh38
rs764933813 Chr.11: 1557847-1557852 on Build GRCh38
rs763720271 Chr.11: 1557770-1557770 on Build GRCh38
rs546651148 Chr.11: 1557819-1557819 on Build GRCh38
rs574421182 Chr.11: 1557740-1557740 on Build GRCh38
rs749311222 Chr.11: 1557715-1557715 on Build GRCh38
rs746039638 Chr.11: 1557809-1557809 on Build GRCh38
rs747250242 Chr.11: 1557752-1557752 on Build GRCh38
rs776448506 Chr.11: 1557754-1557754 on Build GRCh38
rs749215329 Chr.11: 1557821-1557821 on Build GRCh38
rs563099635 Chr.11: 1557757-1557757 on Build GRCh38
rs55763357 Chr.11: 1557719-1557719 on Build GRCh38
rs373838996 Chr.11: 1557776-1557776 on Build GRCh38
rs777768655 Chr.11: 1557743-1557743 on Build GRCh38
rs144147970 Chr.11: 1557853-1557853 on Build GRCh38
rs575190569 Chr.11: 1557763-1557763 on Build GRCh38
rs768686376 Chr.11: 1557826-1557826 on Build GRCh38
rs370107291 Chr.11: 1557814-1557814 on Build GRCh38
rs777858702 Chr.11: 1557799-1557799 on Build GRCh38
rs992830929 Chr.11: 1557714-1557714 on Build GRCh38
rs576750165 Chr.11: 1557662-1557662 on Build GRCh38
rs761392997 Chr.11: 1557841-1557841 on Build GRCh38
rs767343651 Chr.11: 1557847-1557847 on Build GRCh38
rs200008161 Chr.11: 1557773-1557773 on Build GRCh38
rs917297083 Chr.11: 1557739-1557739 on Build GRCh38
rs77411787 Chr.11: 1557802-1557802 on Build GRCh38
rs369338661 Chr.11: 1557707-1557707 on Build GRCh38
rs754286469 Chr.11: 1557774-1557774 on Build GRCh38
rs773959979 Chr.11: 1557772-1557772 on Build GRCh38
rs533179997 Chr.11: 1557753-1557753 on Build GRCh38

View Full Product Details