Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 91214271-91214544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUH
Entrez Gene ID
549
Gene Name
AU RNA binding protein/enoyl-CoA hydratase
Gene Aliases
-
UniGene
Hs.175905
Gene Chromosome Location
Chr.9: 91213815-91361969 on Build GRCh38
Transcripts
NM_001306190.1 NM_001698.2 XM_005252066.3 XM_005252067.4 XM_006717150.3 XM_011518802.2 XM_017014849.1 XM_017014850.1 XM_017014851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467770 c.1003C>T p.R335C Chr.9: 91214365-91214365 on Build GRCh38
4664743 c.997C>T p.P333S Chr.9: 91214371-91214371 on Build GRCh38
1158124 c.996G>T p.R332S Chr.9: 91214372-91214372 on Build GRCh38
4456003 c.986A>T p.K329I Chr.9: 91214382-91214382 on Build GRCh38
6608831 c.946A>G p.I316V Chr.9: 91214422-91214422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760844017 Chr.9: 91214334-91214334 on Build GRCh38
rs552134551 Chr.9: 91214484-91214484 on Build GRCh38
rs774134680 Chr.9: 91214326-91214330 on Build GRCh38
rs766609298 Chr.9: 91214338-91214339 on Build GRCh38
rs751163025 Chr.9: 91214390-91214390 on Build GRCh38
rs748612122 Chr.9: 91214313-91214313 on Build GRCh38
rs773815874 Chr.9: 91214328-91214328 on Build GRCh38
rs563107892 Chr.9: 91214383-91214383 on Build GRCh38
rs771146874 Chr.9: 91214441-91214441 on Build GRCh38
rs529693736 Chr.9: 91214364-91214364 on Build GRCh38
rs766829349 Chr.9: 91214341-91214341 on Build GRCh38
rs778134028 Chr.9: 91214431-91214431 on Build GRCh38
rs987385743 Chr.9: 91214375-91214375 on Build GRCh38
rs924538270 Chr.9: 91214448-91214448 on Build GRCh38
rs946452106 Chr.9: 91214505-91214505 on Build GRCh38
rs754108097 Chr.9: 91214384-91214384 on Build GRCh38
rs774644954 Chr.9: 91214359-91214359 on Build GRCh38
rs755053796 Chr.9: 91214454-91214454 on Build GRCh38
rs765064728 Chr.9: 91214371-91214371 on Build GRCh38
rs754656619 Chr.9: 91214404-91214404 on Build GRCh38
rs964265074 Chr.9: 91214398-91214398 on Build GRCh38
rs747631104 Chr.9: 91214419-91214419 on Build GRCh38
rs1054221227 Chr.9: 91214506-91214506 on Build GRCh38
rs528387187 Chr.9: 91214489-91214489 on Build GRCh38
rs758124622 Chr.9: 91214378-91214378 on Build GRCh38
rs530710210 Chr.9: 91214417-91214417 on Build GRCh38
rs767795059 Chr.9: 91214322-91214323 on Build GRCh38
rs763212178 Chr.9: 91214331-91214331 on Build GRCh38
rs111906408 Chr.9: 91214329-91214329 on Build GRCh38
rs772918339 Chr.9: 91214365-91214365 on Build GRCh38
rs975238186 Chr.9: 91214342-91214342 on Build GRCh38
rs369285903 Chr.9: 91214300-91214300 on Build GRCh38
rs769401148 Chr.9: 91214425-91214425 on Build GRCh38
rs763815758 Chr.9: 91214324-91214325 on Build GRCh38
rs730880312 Chr.9: 91214427-91214427 on Build GRCh38
rs200499826 Chr.9: 91214319-91214319 on Build GRCh38
rs752858725 Chr.9: 91214369-91214369 on Build GRCh38
rs963809756 Chr.9: 91214324-91214324 on Build GRCh38
rs184037865 Chr.9: 91214488-91214488 on Build GRCh38
rs774882099 Chr.9: 91214450-91214450 on Build GRCh38
rs749803389 Chr.9: 91214432-91214432 on Build GRCh38
rs541546111 Chr.9: 91214370-91214370 on Build GRCh38
rs387906757 Chr.9: 91214377-91214377 on Build GRCh38

View Full Product Details