Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85799250-85799483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC8
Entrez Gene ID
10328
Gene Name
ER membrane protein complex subunit 8
Gene Aliases
C16orf2, C16orf4, COX4NB, FAM158B, NOC4
UniGene
Hs.173162
Gene Chromosome Location
Chr.16: 85778624-85799744 on Build GRCh38
Transcripts
NM_001142288.1 NM_006067.4 XM_011522813.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106041 c.12G>A p.V4V Chr.16: 85799284-85799284 on Build GRCh38
3513082 c.8G>A p.G3E Chr.16: 85799288-85799288 on Build GRCh38
2921320 c.20C>T p.T7I Chr.16: 85799276-85799276 on Build GRCh38
974526 c.33C>T p.Y11Y Chr.16: 85799263-85799263 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs890541349 Chr.16: 85799423-85799423 on Build GRCh38
rs753062638 Chr.16: 85799327-85799327 on Build GRCh38
rs993003745 Chr.16: 85799464-85799464 on Build GRCh38
rs769707012 Chr.16: 85799289-85799289 on Build GRCh38
rs975257992 Chr.16: 85799388-85799388 on Build GRCh38
rs757327852 Chr.16: 85799347-85799347 on Build GRCh38
rs530309436 Chr.16: 85799308-85799308 on Build GRCh38
rs926708612 Chr.16: 85799450-85799450 on Build GRCh38
rs112415179 Chr.16: 85799314-85799314 on Build GRCh38
rs1031843427 Chr.16: 85799444-85799444 on Build GRCh38
rs909827100 Chr.16: 85799370-85799370 on Build GRCh38
rs745840387 Chr.16: 85799275-85799275 on Build GRCh38
rs992950492 Chr.16: 85799316-85799316 on Build GRCh38
rs765575648 Chr.16: 85799319-85799319 on Build GRCh38
rs1051880336 Chr.16: 85799418-85799418 on Build GRCh38
rs150142897 Chr.16: 85799292-85799292 on Build GRCh38
rs941319672 Chr.16: 85799432-85799432 on Build GRCh38
rs371404018 Chr.16: 85799304-85799304 on Build GRCh38
rs760105835 Chr.16: 85799309-85799309 on Build GRCh38
rs768360785 Chr.16: 85799299-85799299 on Build GRCh38
rs964251466 Chr.16: 85799387-85799387 on Build GRCh38
rs201630555 Chr.16: 85799330-85799330 on Build GRCh38
rs762635117 Chr.16: 85799298-85799298 on Build GRCh38
rs1017606357 Chr.16: 85799430-85799430 on Build GRCh38
rs2233450 Chr.16: 85799427-85799427 on Build GRCh38
rs979736656 Chr.16: 85799349-85799349 on Build GRCh38
rs956369604 Chr.16: 85799454-85799454 on Build GRCh38
rs966976473 Chr.16: 85799332-85799332 on Build GRCh38
rs761408632 Chr.16: 85799303-85799303 on Build GRCh38
rs773695111 Chr.16: 85799302-85799302 on Build GRCh38
rs904791420 Chr.16: 85799440-85799440 on Build GRCh38
rs371402997 Chr.16: 85799411-85799411 on Build GRCh38
rs938217792 Chr.16: 85799451-85799451 on Build GRCh38
rs896941471 Chr.16: 85799393-85799393 on Build GRCh38
rs751773705 Chr.16: 85799346-85799346 on Build GRCh38
rs1000834522 Chr.16: 85799441-85799441 on Build GRCh38
rs534642187 Chr.16: 85799453-85799453 on Build GRCh38
rs922552050 Chr.16: 85799402-85799402 on Build GRCh38
rs563376246 Chr.16: 85799376-85799376 on Build GRCh38
rs532217840 Chr.16: 85799384-85799384 on Build GRCh38
rs941244989 Chr.16: 85799379-85799379 on Build GRCh38
rs1032229646 Chr.16: 85799286-85799286 on Build GRCh38
rs955836972 Chr.16: 85799297-85799297 on Build GRCh38

View Full Product Details