Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 125124632-125124887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AACS
Entrez Gene ID
65985
Gene Name
acetoacetyl-CoA synthetase
Gene Aliases
ACSF1, SUR-5
UniGene
Hs.656073
Gene Chromosome Location
Chr.12: 125065367-125143333 on Build GRCh38
Transcripts
NM_001319839.1 NM_001319840.1 NM_023928.4 XM_011538692.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
324604 c.1126A>G p.T376A Chr.12: 125124709-125124709 on Build GRCh38
4515580 c.1183_1184CC>TT p.P395L Chr.12: 125124766-125124767 on Build GRCh38
7144194 c.1125C>G p.I375M Chr.12: 125124708-125124708 on Build GRCh38
1666218 c.1122-9G>A p.? Chr.12: 125124696-125124696 on Build GRCh38
5937517 c.1183C>T p.P395S Chr.12: 125124766-125124766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116873665 Chr.12: 125124699-125124699 on Build GRCh38
rs752116179 Chr.12: 125124727-125124727 on Build GRCh38
rs749034583 Chr.12: 125124661-125124661 on Build GRCh38
rs369714366 Chr.12: 125124662-125124662 on Build GRCh38
rs992007510 Chr.12: 125124671-125124671 on Build GRCh38
rs568583813 Chr.12: 125124815-125124815 on Build GRCh38
rs752729710 Chr.12: 125124712-125124712 on Build GRCh38
rs761440706 Chr.12: 125124675-125124675 on Build GRCh38
rs947140229 Chr.12: 125124728-125124728 on Build GRCh38
rs927077550 Chr.12: 125124779-125124779 on Build GRCh38
rs747605076 Chr.12: 125124772-125124772 on Build GRCh38
rs772443589 Chr.12: 125124845-125124845 on Build GRCh38
rs765428730 Chr.12: 125124820-125124820 on Build GRCh38
rs566200830 Chr.12: 125124836-125124836 on Build GRCh38
rs886642589 Chr.12: 125124840-125124840 on Build GRCh38
rs770613745 Chr.12: 125124800-125124800 on Build GRCh38
rs757556585 Chr.12: 125124732-125124732 on Build GRCh38
rs144854520 Chr.12: 125124844-125124844 on Build GRCh38
rs781649055 Chr.12: 125124734-125124734 on Build GRCh38
rs769254610 Chr.12: 125124706-125124706 on Build GRCh38
rs773297852 Chr.12: 125124692-125124692 on Build GRCh38
rs771767542 Chr.12: 125124786-125124786 on Build GRCh38
rs1006458609 Chr.12: 125124843-125124843 on Build GRCh38
rs200498363 Chr.12: 125124696-125124696 on Build GRCh38
rs771295766 Chr.12: 125124660-125124660 on Build GRCh38
rs372637881 Chr.12: 125124792-125124792 on Build GRCh38
rs148397832 Chr.12: 125124753-125124753 on Build GRCh38
rs747305290 Chr.12: 125124658-125124658 on Build GRCh38
rs554211559 Chr.12: 125124832-125124832 on Build GRCh38
rs371499128 Chr.12: 125124809-125124809 on Build GRCh38
rs754609142 Chr.12: 125124757-125124757 on Build GRCh38
rs762206346 Chr.12: 125124846-125124846 on Build GRCh38
rs746985947 Chr.12: 125124794-125124794 on Build GRCh38
rs544839094 Chr.12: 125124834-125124834 on Build GRCh38
rs202075728 Chr.12: 125124691-125124691 on Build GRCh38
rs778192640 Chr.12: 125124722-125124722 on Build GRCh38
rs370284739 Chr.12: 125124768-125124768 on Build GRCh38
rs149638574 Chr.12: 125124714-125124714 on Build GRCh38
rs775045513 Chr.12: 125124709-125124709 on Build GRCh38
rs201676071 Chr.12: 125124682-125124682 on Build GRCh38
rs964657513 Chr.12: 125124672-125124672 on Build GRCh38
rs147704350 Chr.12: 125124826-125124826 on Build GRCh38
rs71447035 Chr.12: 125124681-125124681 on Build GRCh38
rs2291249 Chr.12: 125124838-125124838 on Build GRCh38
rs144461434 Chr.12: 125124744-125124744 on Build GRCh38

View Full Product Details