Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42563208-42563433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COASY
Entrez Gene ID
80347
Gene Name
Coenzyme A synthase
Gene Aliases
DPCK, NBIA6, NBP, PPAT, UKR1, pOV-2
UniGene
Hs.296422
Gene Chromosome Location
Chr.17: 42562074-42566281 on Build GRCh38
Transcripts
NM_001042529.2 NM_001042532.3 NM_025233.6 XM_006722116.3 XM_011525300.1 XM_017025167.1 XM_017025168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
979529 c.590G>A p.G197D Chr.17: 42563212-42563212 on Build GRCh38
5747854 c.653C>T p.A218V Chr.17: 42563275-42563275 on Build GRCh38
1201727 c.604C>T p.R202C Chr.17: 42563226-42563226 on Build GRCh38
5747853 c.740C>T p.A247V Chr.17: 42563275-42563275 on Build GRCh38
3819642 c.726C>T p.V242V Chr.17: 42563261-42563261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137943833 Chr.17: 42563228-42563228 on Build GRCh38
rs375318918 Chr.17: 42563348-42563348 on Build GRCh38
rs761347168 Chr.17: 42563226-42563226 on Build GRCh38
rs780060015 Chr.17: 42563265-42563265 on Build GRCh38
rs376690899 Chr.17: 42563363-42563363 on Build GRCh38
rs774007203 Chr.17: 42563333-42563333 on Build GRCh38
rs767178238 Chr.17: 42563349-42563349 on Build GRCh38
rs770990218 Chr.17: 42563293-42563293 on Build GRCh38
rs1037958091 Chr.17: 42563282-42563282 on Build GRCh38
rs376040856 Chr.17: 42563330-42563330 on Build GRCh38
rs568769350 Chr.17: 42563310-42563310 on Build GRCh38
rs756088989 Chr.17: 42563248-42563248 on Build GRCh38
rs763314347 Chr.17: 42563325-42563325 on Build GRCh38
rs926743163 Chr.17: 42563251-42563251 on Build GRCh38
rs978753509 Chr.17: 42563377-42563377 on Build GRCh38
rs748907041 Chr.17: 42563307-42563308 on Build GRCh38
rs781225303 Chr.17: 42563295-42563295 on Build GRCh38
rs751039293 Chr.17: 42563225-42563225 on Build GRCh38
rs770274030 Chr.17: 42563304-42563304 on Build GRCh38
rs777983479 Chr.17: 42563279-42563279 on Build GRCh38
rs758562645 Chr.17: 42563275-42563275 on Build GRCh38
rs993634231 Chr.17: 42563329-42563329 on Build GRCh38
rs775768555 Chr.17: 42563389-42563389 on Build GRCh38
rs915524540 Chr.17: 42563336-42563336 on Build GRCh38
rs899417712 Chr.17: 42563296-42563296 on Build GRCh38
rs149475124 Chr.17: 42563237-42563237 on Build GRCh38
rs770407833 Chr.17: 42563321-42563324 on Build GRCh38
rs369449670 Chr.17: 42563257-42563257 on Build GRCh38
rs746412403 Chr.17: 42563297-42563297 on Build GRCh38
rs767698305 Chr.17: 42563280-42563280 on Build GRCh38
rs372394606 Chr.17: 42563266-42563266 on Build GRCh38

View Full Product Details