Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112248748-112249021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLET1
Entrez Gene ID
349633
Gene Name
placenta expressed transcript 1
Gene Aliases
C11orf34
UniGene
Hs.632097
Gene Chromosome Location
Chr.11: 112248153-112260860 on Build GRCh38
Transcripts
NM_001145024.1

SNP Detail

SNP ID Chromosome Location
rs907303683 Chr.11: 112248987-112248987 on Build GRCh38
rs146338971 Chr.11: 112249001-112249001 on Build GRCh38
rs994640054 Chr.11: 112248910-112248910 on Build GRCh38
rs181513880 Chr.11: 112248848-112248848 on Build GRCh38
rs749567090 Chr.11: 112248999-112248999 on Build GRCh38
rs1008962522 Chr.11: 112248894-112248894 on Build GRCh38
rs894514646 Chr.11: 112248928-112248928 on Build GRCh38
rs755895566 Chr.11: 112248793-112248793 on Build GRCh38
rs748006216 Chr.11: 112248944-112248944 on Build GRCh38
rs34524729 Chr.11: 112248814-112248815 on Build GRCh38
rs769515781 Chr.11: 112248984-112248984 on Build GRCh38
rs527281354 Chr.11: 112248918-112248918 on Build GRCh38
rs373254138 Chr.11: 112248888-112248888 on Build GRCh38
rs773034925 Chr.11: 112248996-112248996 on Build GRCh38
rs946213032 Chr.11: 112248845-112248845 on Build GRCh38
rs527244689 Chr.11: 112248914-112248914 on Build GRCh38
rs7128270 Chr.11: 112248929-112248929 on Build GRCh38
rs560617061 Chr.11: 112248998-112248998 on Build GRCh38
rs939740098 Chr.11: 112248842-112248842 on Build GRCh38
rs999332147 Chr.11: 112248997-112248997 on Build GRCh38
rs546102505 Chr.11: 112248882-112248882 on Build GRCh38
rs1043217714 Chr.11: 112248847-112248847 on Build GRCh38
rs969192288 Chr.11: 112248993-112248993 on Build GRCh38
rs28548361 Chr.11: 112248884-112248884 on Build GRCh38
rs897543860 Chr.11: 112248906-112248906 on Build GRCh38
rs1014695735 Chr.11: 112248896-112248896 on Build GRCh38
rs912312436 Chr.11: 112248773-112248773 on Build GRCh38
rs755322850 Chr.11: 112248963-112248964 on Build GRCh38

View Full Product Details