Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61309729-61310000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB1
Entrez Gene ID
1642
Gene Name
damage specific DNA binding protein 1
Gene Aliases
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF
UniGene
Hs.290758
Gene Chromosome Location
Chr.11: 61299447-61333212 on Build GRCh38
Transcripts
NM_001923.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4995010 c.2515G>T p.E839* Chr.11: 61309847-61309847 on Build GRCh38
6251766 c.2561C>A p.S854* Chr.11: 61309801-61309801 on Build GRCh38
4516942 c.2547_2548GG>AA p.V850I Chr.11: 61309814-61309815 on Build GRCh38
6588443 c.2556G>C p.Q852H Chr.11: 61309806-61309806 on Build GRCh38
1746373 c.2437G>A p.A813T Chr.11: 61309925-61309925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200880349 Chr.11: 61309960-61309960 on Build GRCh38
rs779557754 Chr.11: 61309784-61309784 on Build GRCh38
rs145122902 Chr.11: 61309757-61309757 on Build GRCh38
rs779271195 Chr.11: 61309944-61309944 on Build GRCh38
rs764196921 Chr.11: 61309761-61309761 on Build GRCh38
rs759340755 Chr.11: 61309965-61309969 on Build GRCh38
rs114169860 Chr.11: 61309769-61309769 on Build GRCh38
rs146448474 Chr.11: 61309854-61309854 on Build GRCh38
rs775572938 Chr.11: 61309819-61309819 on Build GRCh38
rs1052471660 Chr.11: 61309805-61309805 on Build GRCh38
rs778127067 Chr.11: 61309944-61309954 on Build GRCh38
rs760853450 Chr.11: 61309823-61309823 on Build GRCh38
rs374720910 Chr.11: 61309969-61309969 on Build GRCh38
rs902122743 Chr.11: 61309945-61309945 on Build GRCh38
rs754574975 Chr.11: 61309901-61309901 on Build GRCh38
rs746594152 Chr.11: 61309785-61309785 on Build GRCh38
rs749814416 Chr.11: 61309790-61309790 on Build GRCh38
rs367819676 Chr.11: 61309873-61309873 on Build GRCh38
rs746313813 Chr.11: 61309959-61309959 on Build GRCh38
rs974593816 Chr.11: 61309876-61309876 on Build GRCh38
rs751149642 Chr.11: 61309899-61309899 on Build GRCh38
rs772398657 Chr.11: 61309817-61309817 on Build GRCh38
rs772295890 Chr.11: 61309964-61309964 on Build GRCh38
rs761326357 Chr.11: 61309976-61309980 on Build GRCh38
rs772471087 Chr.11: 61309941-61309942 on Build GRCh38
rs750246920 Chr.11: 61309874-61309874 on Build GRCh38
rs747149302 Chr.11: 61309958-61309958 on Build GRCh38
rs771064283 Chr.11: 61309963-61309964 on Build GRCh38
rs776641320 Chr.11: 61309963-61309963 on Build GRCh38
rs773926924 Chr.11: 61309974-61309974 on Build GRCh38
rs889304958 Chr.11: 61309864-61309864 on Build GRCh38
rs758122677 Chr.11: 61309893-61309893 on Build GRCh38
rs773867110 Chr.11: 61309970-61309970 on Build GRCh38
rs768122609 Chr.11: 61309786-61309786 on Build GRCh38
rs892471588 Chr.11: 61309807-61309807 on Build GRCh38
rs778873072 Chr.11: 61309771-61309771 on Build GRCh38
rs757772014 Chr.11: 61309941-61309941 on Build GRCh38
rs771383564 Chr.11: 61309800-61309800 on Build GRCh38
rs780707990 Chr.11: 61309920-61309920 on Build GRCh38
rs939235024 Chr.11: 61309801-61309801 on Build GRCh38
rs752833153 Chr.11: 61309749-61309749 on Build GRCh38
rs763806910 Chr.11: 61309971-61309972 on Build GRCh38
rs753800101 Chr.11: 61309763-61309763 on Build GRCh38
rs907612805 Chr.11: 61309973-61309973 on Build GRCh38
rs775833977 Chr.11: 61309967-61309967 on Build GRCh38
rs768263767 Chr.11: 61309933-61309933 on Build GRCh38
rs769743113 Chr.11: 61309966-61309966 on Build GRCh38
rs759759255 Chr.11: 61309816-61309816 on Build GRCh38
rs778300920 Chr.11: 61309787-61309787 on Build GRCh38
rs775200461 Chr.11: 61309968-61309969 on Build GRCh38
rs1008118392 Chr.11: 61309913-61309913 on Build GRCh38
rs762727617 Chr.11: 61309970-61309971 on Build GRCh38
rs201599936 Chr.11: 61309859-61309859 on Build GRCh38
rs372773947 Chr.11: 61309898-61309898 on Build GRCh38
rs774780663 Chr.11: 61309815-61309815 on Build GRCh38
rs764250253 Chr.11: 61309848-61309848 on Build GRCh38
rs866389161 Chr.11: 61309857-61309857 on Build GRCh38
rs371631979 Chr.11: 61309778-61309778 on Build GRCh38
rs750146956 Chr.11: 61309776-61309776 on Build GRCh38

View Full Product Details