Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36406114-36406375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANLN
Entrez Gene ID
54443
Gene Name
anillin actin binding protein
Gene Aliases
FSGS8, Scraps, scra
UniGene
Hs.62180
Gene Chromosome Location
Chr.7: 36389803-36453791 on Build GRCh38
Transcripts
NM_001284301.2 NM_001284302.2 NM_018685.4 XM_006715746.1 XM_006715747.3 XM_017012354.1 XM_017012355.1 XM_017012356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
318698 c.601C>G p.R201G Chr.7: 36406294-36406294 on Build GRCh38
3929035 c.518C>A p.P173Q Chr.7: 36406211-36406211 on Build GRCh38
4409917 c.544T>C p.S182P Chr.7: 36406237-36406237 on Build GRCh38
5026740 c.559C>G p.L187V Chr.7: 36406252-36406252 on Build GRCh38
6177526 c.585A>T p.P195P Chr.7: 36406278-36406278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749987420 Chr.7: 36406240-36406240 on Build GRCh38
rs61737563 Chr.7: 36406246-36406246 on Build GRCh38
rs200557457 Chr.7: 36406330-36406330 on Build GRCh38
rs767242076 Chr.7: 36406318-36406318 on Build GRCh38
rs753136477 Chr.7: 36406343-36406343 on Build GRCh38
rs747193872 Chr.7: 36406258-36406258 on Build GRCh38
rs140160802 Chr.7: 36406291-36406291 on Build GRCh38
rs770850733 Chr.7: 36406149-36406149 on Build GRCh38
rs369406062 Chr.7: 36406325-36406325 on Build GRCh38
rs28657426 Chr.7: 36406225-36406225 on Build GRCh38
rs755553777 Chr.7: 36406162-36406162 on Build GRCh38
rs1031713784 Chr.7: 36406155-36406155 on Build GRCh38
rs369115782 Chr.7: 36406165-36406165 on Build GRCh38
rs555173750 Chr.7: 36406232-36406232 on Build GRCh38
rs1040608350 Chr.7: 36406147-36406147 on Build GRCh38
rs543090284 Chr.7: 36406290-36406290 on Build GRCh38
rs773805253 Chr.7: 36406238-36406238 on Build GRCh38
rs770014887 Chr.7: 36406285-36406285 on Build GRCh38
rs148482760 Chr.7: 36406217-36406217 on Build GRCh38
rs528529344 Chr.7: 36406336-36406336 on Build GRCh38
rs139482132 Chr.7: 36406268-36406268 on Build GRCh38
rs557429446 Chr.7: 36406248-36406248 on Build GRCh38
rs760132700 Chr.7: 36406335-36406335 on Build GRCh38
rs755450565 Chr.7: 36406241-36406241 on Build GRCh38
rs775599772 Chr.7: 36406173-36406173 on Build GRCh38
rs866710441 Chr.7: 36406338-36406338 on Build GRCh38
rs748967747 Chr.7: 36406239-36406239 on Build GRCh38
rs761347630 Chr.7: 36406304-36406304 on Build GRCh38
rs775849590 Chr.7: 36406274-36406274 on Build GRCh38
rs377576846 Chr.7: 36406315-36406315 on Build GRCh38
rs972370199 Chr.7: 36406313-36406313 on Build GRCh38
rs373587417 Chr.7: 36406295-36406295 on Build GRCh38
rs771815300 Chr.7: 36406151-36406151 on Build GRCh38
rs776838798 Chr.7: 36406152-36406152 on Build GRCh38
rs197367 Chr.7: 36406247-36406247 on Build GRCh38
rs376264665 Chr.7: 36406269-36406269 on Build GRCh38
rs386712160 Chr.7: 36406246-36406247 on Build GRCh38
rs902125007 Chr.7: 36406150-36406150 on Build GRCh38
rs147807983 Chr.7: 36406296-36406296 on Build GRCh38

View Full Product Details