Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46643113-46643382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPAF1
Entrez Gene ID
64756
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Gene Aliases
ATP11, ATP11p
UniGene
Hs.100874
Gene Chromosome Location
Chr.1: 46635039-46668427 on Build GRCh38
Transcripts
NM_001042546.2 NM_001243728.1 NM_001256418.1 NM_022745.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6460531 c.717T>A p.I239I Chr.1: 46643269-46643269 on Build GRCh38
3865688 c.766A>G p.T256A Chr.1: 46643220-46643220 on Build GRCh38
3805457 c.743A>G p.E248G Chr.1: 46643243-46643243 on Build GRCh38
2191384 c.689G>A p.R230Q Chr.1: 46643297-46643297 on Build GRCh38
2191383 c.704C>T p.A235V Chr.1: 46643282-46643282 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs367657762 Chr.1: 46643298-46643298 on Build GRCh38
rs758105632 Chr.1: 46643311-46643311 on Build GRCh38
rs904012563 Chr.1: 46643307-46643307 on Build GRCh38
rs536871523 Chr.1: 46643345-46643345 on Build GRCh38
rs569305969 Chr.1: 46643275-46643275 on Build GRCh38
rs765373799 Chr.1: 46643242-46643242 on Build GRCh38
rs780911565 Chr.1: 46643174-46643174 on Build GRCh38
rs374884433 Chr.1: 46643348-46643348 on Build GRCh38
rs745838000 Chr.1: 46643353-46643353 on Build GRCh38
rs979608413 Chr.1: 46643303-46643303 on Build GRCh38
rs200333485 Chr.1: 46643200-46643200 on Build GRCh38
rs763191064 Chr.1: 46643263-46643263 on Build GRCh38
rs1027213390 Chr.1: 46643140-46643140 on Build GRCh38
rs58805974 Chr.1: 46643157-46643157 on Build GRCh38
rs551298709 Chr.1: 46643206-46643206 on Build GRCh38
rs776698866 Chr.1: 46643226-46643226 on Build GRCh38
rs754433499 Chr.1: 46643167-46643170 on Build GRCh38
rs754580580 Chr.1: 46643160-46643160 on Build GRCh38
rs201098574 Chr.1: 46643187-46643187 on Build GRCh38
rs371773861 Chr.1: 46643335-46643335 on Build GRCh38
rs368720539 Chr.1: 46643230-46643230 on Build GRCh38
rs371462772 Chr.1: 46643292-46643292 on Build GRCh38
rs757673808 Chr.1: 46643296-46643296 on Build GRCh38
rs778360613 Chr.1: 46643178-46643178 on Build GRCh38
rs923494405 Chr.1: 46643232-46643232 on Build GRCh38
rs187654168 Chr.1: 46643352-46643352 on Build GRCh38
rs757729248 Chr.1: 46643155-46643155 on Build GRCh38
rs736188 Chr.1: 46643156-46643156 on Build GRCh38
rs375918153 Chr.1: 46643297-46643297 on Build GRCh38
rs778518808 Chr.1: 46643308-46643308 on Build GRCh38
rs775525603 Chr.1: 46643261-46643261 on Build GRCh38
rs751099506 Chr.1: 46643156-46643165 on Build GRCh38
rs752020559 Chr.1: 46643144-46643144 on Build GRCh38
rs747688130 Chr.1: 46643237-46643237 on Build GRCh38
rs377071749 Chr.1: 46643146-46643146 on Build GRCh38
rs757491853 Chr.1: 46643253-46643253 on Build GRCh38
rs376154299 Chr.1: 46643220-46643220 on Build GRCh38
rs747654069 Chr.1: 46643310-46643310 on Build GRCh38

View Full Product Details