Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 246621234-246621495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNST
Entrez Gene ID
163882
Gene Name
consortin, connexin sorting protein
Gene Aliases
C1orf71, PPP1R64
UniGene
Hs.368353
Gene Chromosome Location
Chr.1: 246566337-246668587 on Build GRCh38
Transcripts
NM_001139459.1 NM_152609.2 XM_005273081.3 XM_005273083.4 XM_011544110.2 XM_011544111.1 XM_011544112.1 XM_011544113.1 XM_011544114.2 XM_017000487.1 XM_017000488.1 XM_017000489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4778135 c.428G>A p.S143N Chr.1: 246621477-246621477 on Build GRCh38
1731538 c.383delT p.S130fs*7 Chr.1: 246621432-246621432 on Build GRCh38
4917571 c.424C>T p.L142L Chr.1: 246621473-246621473 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781653180 Chr.1: 246621404-246621404 on Build GRCh38
rs1000176466 Chr.1: 246621270-246621270 on Build GRCh38
rs763293106 Chr.1: 246621378-246621378 on Build GRCh38
rs761637006 Chr.1: 246621460-246621460 on Build GRCh38
rs530164942 Chr.1: 246621369-246621369 on Build GRCh38
rs561202082 Chr.1: 246621353-246621353 on Build GRCh38
rs765158361 Chr.1: 246621462-246621462 on Build GRCh38
rs772207995 Chr.1: 246621456-246621456 on Build GRCh38
rs765735549 Chr.1: 246621282-246621282 on Build GRCh38
rs751059668 Chr.1: 246621473-246621473 on Build GRCh38
rs1006090800 Chr.1: 246621392-246621392 on Build GRCh38
rs973059444 Chr.1: 246621453-246621453 on Build GRCh38
rs544662694 Chr.1: 246621293-246621293 on Build GRCh38
rs776388859 Chr.1: 246621459-246621459 on Build GRCh38
rs75636113 Chr.1: 246621437-246621437 on Build GRCh38
rs773196569 Chr.1: 246621467-246621467 on Build GRCh38
rs998096436 Chr.1: 246621266-246621266 on Build GRCh38
rs778460133 Chr.1: 246621420-246621420 on Build GRCh38
rs372687859 Chr.1: 246621439-246621439 on Build GRCh38
rs749323606 Chr.1: 246621434-246621434 on Build GRCh38
rs754593918 Chr.1: 246621395-246621395 on Build GRCh38
rs368913317 Chr.1: 246621412-246621412 on Build GRCh38
rs748543363 Chr.1: 246621407-246621407 on Build GRCh38
rs150125148 Chr.1: 246621373-246621373 on Build GRCh38
rs746033634 Chr.1: 246621447-246621447 on Build GRCh38
rs961248929 Chr.1: 246621289-246621289 on Build GRCh38
rs762815022 Chr.1: 246621471-246621471 on Build GRCh38
rs765850766 Chr.1: 246621472-246621472 on Build GRCh38

View Full Product Details