Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 23693319-23693590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL2
Entrez Gene ID
1993
Gene Name
ELAV like neuron-specific RNA binding protein 2
Gene Aliases
HEL-N1, HELN1, HUB
UniGene
Hs.166109
Gene Chromosome Location
Chr.9: 23690099-23850757 on Build GRCh38
Transcripts
NM_001171195.1 NM_001171197.1 NM_004432.3 XM_005251393.3 XM_005251394.3 XM_005251395.3 XM_006716734.3 XM_006716735.3 XM_006716736.3 XM_011517774.2 XM_011517776.2 XM_011517777.2 XM_011517778.2 XM_011517779.2 XM_011517780.2 XM_011517783.2 XM_011517784.2 XM_011517785.2 XM_011517786.2 XM_017014408.1 XM_017014409.1 XM_017014410.1 XM_017014411.1 XM_017014412.1 XM_017014413.1 XM_017014414.1 XM_017014415.1 XM_017014416.1 XM_017014417.1 XM_017014418.1 XM_017014419.1 XM_017014420.1 XM_017014421.1 XM_017014422.1 XM_017014423.1 XM_017014424.1 XM_017014425.1 XM_017014426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432669 c.813C>G p.L271L Chr.9: 23693471-23693471 on Build GRCh38
6183423 c.742G>A p.G248R Chr.9: 23693458-23693458 on Build GRCh38
5005233 c.820G>A p.A274T Chr.9: 23693464-23693464 on Build GRCh38
6434398 c.835A>T p.R279W Chr.9: 23693449-23693449 on Build GRCh38
5005232 c.736G>A p.A246T Chr.9: 23693464-23693464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184358313 Chr.9: 23693403-23693403 on Build GRCh38
rs569757223 Chr.9: 23693533-23693533 on Build GRCh38
rs749915789 Chr.9: 23693418-23693418 on Build GRCh38
rs761514986 Chr.9: 23693414-23693414 on Build GRCh38
rs752363231 Chr.9: 23693372-23693372 on Build GRCh38
rs200428427 Chr.9: 23693415-23693415 on Build GRCh38
rs370429986 Chr.9: 23693550-23693550 on Build GRCh38
rs1030761230 Chr.9: 23693538-23693538 on Build GRCh38
rs755650566 Chr.9: 23693421-23693421 on Build GRCh38
rs773133788 Chr.9: 23693445-23693455 on Build GRCh38
rs989372996 Chr.9: 23693564-23693564 on Build GRCh38
rs775502029 Chr.9: 23693459-23693459 on Build GRCh38
rs758691628 Chr.9: 23693430-23693430 on Build GRCh38
rs757476671 Chr.9: 23693441-23693441 on Build GRCh38
rs761467484 Chr.9: 23693412-23693412 on Build GRCh38
rs567806256 Chr.9: 23693361-23693361 on Build GRCh38
rs767476313 Chr.9: 23693439-23693440 on Build GRCh38
rs888081868 Chr.9: 23693422-23693422 on Build GRCh38
rs747388241 Chr.9: 23693437-23693437 on Build GRCh38
rs1027076068 Chr.9: 23693371-23693371 on Build GRCh38
rs763955294 Chr.9: 23693408-23693408 on Build GRCh38
rs768535058 Chr.9: 23693478-23693478 on Build GRCh38
rs951307120 Chr.9: 23693399-23693399 on Build GRCh38
rs761551993 Chr.9: 23693523-23693523 on Build GRCh38
rs369362020 Chr.9: 23693536-23693536 on Build GRCh38
rs1012452033 Chr.9: 23693423-23693423 on Build GRCh38
rs914594929 Chr.9: 23693567-23693567 on Build GRCh38
rs745897718 Chr.9: 23693447-23693447 on Build GRCh38
rs753215989 Chr.9: 23693428-23693428 on Build GRCh38
rs774081305 Chr.9: 23693409-23693409 on Build GRCh38
rs547583733 Chr.9: 23693348-23693348 on Build GRCh38
rs749233048 Chr.9: 23693460-23693460 on Build GRCh38
rs375638740 Chr.9: 23693529-23693529 on Build GRCh38
rs201078756 Chr.9: 23693528-23693528 on Build GRCh38
rs1034870551 Chr.9: 23693368-23693368 on Build GRCh38
rs778191638 Chr.9: 23693435-23693435 on Build GRCh38
rs770010735 Chr.9: 23693457-23693457 on Build GRCh38
rs372344586 Chr.9: 23693499-23693499 on Build GRCh38
rs765683959 Chr.9: 23693425-23693425 on Build GRCh38

View Full Product Details