Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31657951-31658224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOM
Entrez Gene ID
55937
Gene Name
apolipoprotein M
Gene Aliases
G3a, HSPC336, NG20, apo-M
UniGene
Hs.534468
Gene Chromosome Location
Chr.6: 31652410-31658210 on Build GRCh38
Transcripts
NM_001256169.1 NM_019101.2 NR_045828.1 XM_006715150.3 XM_017011279.1

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
UniGene
Hs.440900
Gene Chromosome Location
Chr.6: 31639028-31660900 on Build GRCh38
Transcripts
NM_001256169.1 NM_019101.2 NR_045828.1 XM_006715150.3 XM_017011279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3394092 c.542-1G>C p.? Chr.6: 31658063-31658063 on Build GRCh38
6173642 c.566G>A p.*189* Chr.6: 31658088-31658088 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs946613827 Chr.6: 31658193-31658193 on Build GRCh38
rs760614075 Chr.6: 31658162-31658162 on Build GRCh38
rs772787234 Chr.6: 31658059-31658060 on Build GRCh38
rs745781412 Chr.6: 31658042-31658042 on Build GRCh38
rs757585801 Chr.6: 31658086-31658086 on Build GRCh38
rs959197755 Chr.6: 31658135-31658135 on Build GRCh38
rs1015252531 Chr.6: 31658012-31658012 on Build GRCh38
rs1003859566 Chr.6: 31657993-31657993 on Build GRCh38
rs916284431 Chr.6: 31658105-31658105 on Build GRCh38
rs200256338 Chr.6: 31658054-31658054 on Build GRCh38
rs371669056 Chr.6: 31658123-31658123 on Build GRCh38
rs992659555 Chr.6: 31658151-31658151 on Build GRCh38
rs749417439 Chr.6: 31658133-31658133 on Build GRCh38
rs28432254 Chr.6: 31658107-31658107 on Build GRCh38
rs1043689928 Chr.6: 31658200-31658200 on Build GRCh38
rs1024985988 Chr.6: 31658190-31658190 on Build GRCh38
rs966699827 Chr.6: 31658198-31658198 on Build GRCh38
rs370445372 Chr.6: 31658122-31658122 on Build GRCh38
rs995429910 Chr.6: 31658106-31658106 on Build GRCh38
rs373041094 Chr.6: 31658042-31658042 on Build GRCh38
rs574907406 Chr.6: 31658176-31658176 on Build GRCh38
rs540556905 Chr.6: 31658099-31658099 on Build GRCh38
rs772129390 Chr.6: 31658139-31658139 on Build GRCh38
rs898445650 Chr.6: 31658103-31658103 on Build GRCh38

View Full Product Details