Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83049838-83050081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS4
Entrez Gene ID
51138
Gene Name
COP9 signalosome subunit 4
Gene Aliases
CSN4
UniGene
Hs.190384
Gene Chromosome Location
Chr.4: 83035086-83075818 on Build GRCh38
Transcripts
NM_001258006.1 NM_016129.2 XM_011532019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3606579 c.314C>T p.S105F Chr.4: 83049888-83049888 on Build GRCh38
1486123 c.410+1G>A p.? Chr.4: 83049985-83049985 on Build GRCh38
260258 c.349G>T p.E117* Chr.4: 83049923-83049923 on Build GRCh38
1753856 c.332C>T p.A111V Chr.4: 83049906-83049906 on Build GRCh38
30531 c.385G>C p.G129R Chr.4: 83049959-83049959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776566721 Chr.4: 83049904-83049904 on Build GRCh38
rs767963743 Chr.4: 83050014-83050014 on Build GRCh38
rs767702565 Chr.4: 83049913-83049913 on Build GRCh38
rs372551519 Chr.4: 83050035-83050035 on Build GRCh38
rs546049792 Chr.4: 83050045-83050045 on Build GRCh38
rs891594580 Chr.4: 83049937-83049937 on Build GRCh38
rs767453225 Chr.4: 83049931-83049931 on Build GRCh38
rs958406821 Chr.4: 83050036-83050036 on Build GRCh38
rs376606343 Chr.4: 83050022-83050022 on Build GRCh38
rs752346667 Chr.4: 83050000-83050000 on Build GRCh38
rs759023199 Chr.4: 83049930-83049930 on Build GRCh38
rs550582534 Chr.4: 83050043-83050043 on Build GRCh38
rs146149758 Chr.4: 83049871-83049871 on Build GRCh38
rs1042453849 Chr.4: 83050044-83050044 on Build GRCh38
rs750886172 Chr.4: 83050015-83050015 on Build GRCh38
rs557021764 Chr.4: 83050004-83050004 on Build GRCh38
rs766363793 Chr.4: 83049890-83049890 on Build GRCh38
rs761072865 Chr.4: 83050024-83050024 on Build GRCh38

View Full Product Details