Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128862738-128862995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1F
Entrez Gene ID
9296
Gene Name
ATPase H+ transporting V1 subunit F
Gene Aliases
ATP6S14, VATF, Vma7
UniGene
Hs.78089
Gene Chromosome Location
Chr.7: 128862803-128865849 on Build GRCh38
Transcripts
NM_001198909.1 NM_004231.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1085391 c.80A>T p.E27V Chr.7: 128862984-128862984 on Build GRCh38
318850 c.40G>A p.E14K Chr.7: 128862944-128862944 on Build GRCh38
6176205 c.12G>T p.R4S Chr.7: 128862916-128862916 on Build GRCh38
1131715 c.71G>A p.G24D Chr.7: 128862975-128862975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773505920 Chr.7: 128862892-128862892 on Build GRCh38
rs143547947 Chr.7: 128862915-128862915 on Build GRCh38
rs751485057 Chr.7: 128862898-128862898 on Build GRCh38
rs746947613 Chr.7: 128862857-128862857 on Build GRCh38
rs374713383 Chr.7: 128862895-128862895 on Build GRCh38
rs751026768 Chr.7: 128862944-128862944 on Build GRCh38
rs908459545 Chr.7: 128862894-128862894 on Build GRCh38
rs113816833 Chr.7: 128862893-128862893 on Build GRCh38
rs953314449 Chr.7: 128862831-128862831 on Build GRCh38
rs956715798 Chr.7: 128862812-128862812 on Build GRCh38
rs200605796 Chr.7: 128862899-128862899 on Build GRCh38
rs1019060536 Chr.7: 128862847-128862847 on Build GRCh38
rs761081102 Chr.7: 128862930-128862930 on Build GRCh38
rs368558184 Chr.7: 128862924-128862924 on Build GRCh38
rs376648104 Chr.7: 128862886-128862886 on Build GRCh38
rs892003139 Chr.7: 128862827-128862827 on Build GRCh38
rs1000552330 Chr.7: 128862782-128862782 on Build GRCh38
rs948958405 Chr.7: 128862910-128862910 on Build GRCh38
rs761662267 Chr.7: 128862947-128862947 on Build GRCh38
rs1804887 Chr.7: 128862927-128862927 on Build GRCh38
rs200558362 Chr.7: 128862916-128862916 on Build GRCh38
rs748614632 Chr.7: 128862923-128862923 on Build GRCh38
rs1030726554 Chr.7: 128862786-128862786 on Build GRCh38
rs147175274 Chr.7: 128862918-128862918 on Build GRCh38
rs11547837 Chr.7: 128862866-128862866 on Build GRCh38
rs770085359 Chr.7: 128862874-128862874 on Build GRCh38
rs755969437 Chr.7: 128862909-128862909 on Build GRCh38
rs781003764 Chr.7: 128862869-128862869 on Build GRCh38
rs141175269 Chr.7: 128862932-128862932 on Build GRCh38
rs771259209 Chr.7: 128862931-128862931 on Build GRCh38
rs746118590 Chr.7: 128862870-128862870 on Build GRCh38
rs781641951 Chr.7: 128862785-128862785 on Build GRCh38
rs148608984 Chr.7: 128862928-128862928 on Build GRCh38
rs763537679 Chr.7: 128862943-128862943 on Build GRCh38
rs780287248 Chr.7: 128862880-128862880 on Build GRCh38
rs904171667 Chr.7: 128862779-128862779 on Build GRCh38
rs774760810 Chr.7: 128862890-128862890 on Build GRCh38
rs780481596 Chr.7: 128862912-128862912 on Build GRCh38
rs763633085 Chr.7: 128862896-128862896 on Build GRCh38
rs562393712 Chr.7: 128862806-128862806 on Build GRCh38
rs551221342 Chr.7: 128862853-128862853 on Build GRCh38
rs774350378 Chr.7: 128862891-128862891 on Build GRCh38
rs903555526 Chr.7: 128862758-128862758 on Build GRCh38
rs925660143 Chr.7: 128862925-128862925 on Build GRCh38
rs761957471 Chr.7: 128862933-128862933 on Build GRCh38
rs1030619677 Chr.7: 128862819-128862819 on Build GRCh38
rs879357062 Chr.7: 128862830-128862830 on Build GRCh38
rs750176876 Chr.7: 128862902-128862902 on Build GRCh38
rs535408522 Chr.7: 128862935-128862935 on Build GRCh38
rs986075987 Chr.7: 128862832-128862832 on Build GRCh38
rs117657652 Chr.7: 128862817-128862817 on Build GRCh38
rs767560631 Chr.7: 128862952-128862952 on Build GRCh38
rs371637318 Chr.7: 128862901-128862901 on Build GRCh38

View Full Product Details