Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181053288-181053527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL9
Entrez Gene ID
153579
Gene Name
butyrophilin like 9
Gene Aliases
BTN3, BTN8, VDLS1900
UniGene
Hs.546502
Gene Chromosome Location
Chr.5: 181039875-181061525 on Build GRCh38
Transcripts
NM_001308245.1 NM_152547.4 XM_006714823.3 XM_006714824.3 XM_006714825.3 XM_011534444.2 XM_017009113.1 XM_017009114.1 XM_017009115.1 XM_017009116.1 XM_017009117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4958363 c.842G>A p.R281Q Chr.5: 181053305-181053305 on Build GRCh38
5589854 c.851G>A p.R284Q Chr.5: 181053314-181053314 on Build GRCh38
6547031 c.858G>T p.K286N Chr.5: 181053473-181053473 on Build GRCh38
6547033 c.858G>T p.K286N Chr.5: 181053473-181053473 on Build GRCh38
5589855 c.851G>A p.R284Q Chr.5: 181053314-181053314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767973902 Chr.5: 181053450-181053450 on Build GRCh38
rs750431971 Chr.5: 181053326-181053327 on Build GRCh38
rs768324986 Chr.5: 181053366-181053366 on Build GRCh38
rs1054727695 Chr.5: 181053382-181053382 on Build GRCh38
rs540698898 Chr.5: 181053419-181053419 on Build GRCh38
rs753155365 Chr.5: 181053456-181053456 on Build GRCh38
rs930669905 Chr.5: 181053461-181053461 on Build GRCh38
rs7709523 Chr.5: 181053363-181053363 on Build GRCh38
rs111934728 Chr.5: 181053391-181053391 on Build GRCh38
rs148177643 Chr.5: 181053421-181053421 on Build GRCh38
rs1029884664 Chr.5: 181053399-181053399 on Build GRCh38
rs766929317 Chr.5: 181053305-181053305 on Build GRCh38
rs532902047 Chr.5: 181053438-181053438 on Build GRCh38
rs1046305598 Chr.5: 181053353-181053353 on Build GRCh38
rs990081067 Chr.5: 181053392-181053392 on Build GRCh38
rs930182545 Chr.5: 181053343-181053344 on Build GRCh38
rs957061223 Chr.5: 181053385-181053385 on Build GRCh38
rs752956953 Chr.5: 181053325-181053325 on Build GRCh38
rs202103036 Chr.5: 181053316-181053316 on Build GRCh38
rs757093264 Chr.5: 181053337-181053337 on Build GRCh38
rs939308760 Chr.5: 181053306-181053306 on Build GRCh38
rs745859504 Chr.5: 181053354-181053354 on Build GRCh38
rs1036471607 Chr.5: 181053426-181053426 on Build GRCh38
rs753983857 Chr.5: 181053333-181053333 on Build GRCh38
rs573375473 Chr.5: 181053414-181053414 on Build GRCh38
rs943528296 Chr.5: 181053509-181053509 on Build GRCh38
rs764401743 Chr.5: 181053475-181053475 on Build GRCh38
rs981686278 Chr.5: 181053418-181053418 on Build GRCh38
rs543007670 Chr.5: 181053447-181053447 on Build GRCh38
rs757364253 Chr.5: 181053499-181053499 on Build GRCh38
rs778923657 Chr.5: 181053341-181053341 on Build GRCh38
rs138850635 Chr.5: 181053326-181053326 on Build GRCh38
rs780492903 Chr.5: 181053393-181053393 on Build GRCh38
rs1031287339 Chr.5: 181053373-181053373 on Build GRCh38
rs370413935 Chr.5: 181053323-181053323 on Build GRCh38
rs1036376252 Chr.5: 181053314-181053314 on Build GRCh38
rs759980197 Chr.5: 181053321-181053321 on Build GRCh38
rs753803664 Chr.5: 181053487-181053487 on Build GRCh38
rs889367676 Chr.5: 181053507-181053507 on Build GRCh38
rs985337999 Chr.5: 181053437-181053437 on Build GRCh38
rs774921727 Chr.5: 181053446-181053446 on Build GRCh38
rs1049279567 Chr.5: 181053483-181053483 on Build GRCh38
rs7727802 Chr.5: 181053365-181053365 on Build GRCh38
rs771841279 Chr.5: 181053356-181053356 on Build GRCh38
rs368574604 Chr.5: 181053422-181053422 on Build GRCh38
rs999381646 Chr.5: 181053396-181053396 on Build GRCh38

View Full Product Details