Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99739587-99739836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUTC
Entrez Gene ID
51076
Gene Name
cutC copper transporter
Gene Aliases
CGI-32
UniGene
Hs.16606
Gene Chromosome Location
Chr.10: 99732201-99756137 on Build GRCh38
Transcripts
NM_015960.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4055011 c.174G>A p.G58G Chr.10: 99739750-99739750 on Build GRCh38
7276562 c.176G>A p.G59E Chr.10: 99739752-99739752 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs948554667 Chr.10: 99739648-99739648 on Build GRCh38
rs368135222 Chr.10: 99739800-99739800 on Build GRCh38
rs180812309 Chr.10: 99739749-99739749 on Build GRCh38
rs986545993 Chr.10: 99739623-99739623 on Build GRCh38
rs925234431 Chr.10: 99739646-99739646 on Build GRCh38
rs373698925 Chr.10: 99739719-99739719 on Build GRCh38
rs758861003 Chr.10: 99739686-99739686 on Build GRCh38
rs140301005 Chr.10: 99739718-99739718 on Build GRCh38
rs760653015 Chr.10: 99739661-99739661 on Build GRCh38
rs747030049 Chr.10: 99739731-99739731 on Build GRCh38
rs768446164 Chr.10: 99739706-99739706 on Build GRCh38
rs370921548 Chr.10: 99739663-99739663 on Build GRCh38
rs79582783 Chr.10: 99739791-99739791 on Build GRCh38
rs755576277 Chr.10: 99739693-99739693 on Build GRCh38
rs762712218 Chr.10: 99739747-99739747 on Build GRCh38
rs72832531 Chr.10: 99739756-99739756 on Build GRCh38
rs752147953 Chr.10: 99739706-99739708 on Build GRCh38
rs937370572 Chr.10: 99739710-99739710 on Build GRCh38
rs1017961936 Chr.10: 99739629-99739630 on Build GRCh38
rs778813695 Chr.10: 99739707-99739707 on Build GRCh38
rs998059282 Chr.10: 99739789-99739789 on Build GRCh38
rs776782870 Chr.10: 99739748-99739748 on Build GRCh38
rs770846461 Chr.10: 99739738-99739738 on Build GRCh38
rs1045871841 Chr.10: 99739680-99739680 on Build GRCh38
rs541915589 Chr.10: 99739667-99739667 on Build GRCh38
rs765473294 Chr.10: 99739751-99739751 on Build GRCh38
rs773013737 Chr.10: 99739729-99739729 on Build GRCh38
rs765524593 Chr.10: 99739683-99739683 on Build GRCh38
rs369394173 Chr.10: 99739622-99739622 on Build GRCh38
rs949187746 Chr.10: 99739672-99739672 on Build GRCh38
rs781062722 Chr.10: 99739754-99739754 on Build GRCh38
rs111409102 Chr.10: 99739776-99739776 on Build GRCh38
rs752156713 Chr.10: 99739689-99739689 on Build GRCh38
rs777624737 Chr.10: 99739812-99739812 on Build GRCh38
rs200200786 Chr.10: 99739790-99739790 on Build GRCh38
rs765759117 Chr.10: 99739785-99739785 on Build GRCh38
rs935205964 Chr.10: 99739660-99739660 on Build GRCh38
rs758120496 Chr.10: 99739809-99739809 on Build GRCh38
rs898747392 Chr.10: 99739760-99739760 on Build GRCh38
rs200742740 Chr.10: 99739778-99739778 on Build GRCh38
rs759756346 Chr.10: 99739676-99739676 on Build GRCh38
rs558736025 Chr.10: 99739715-99739715 on Build GRCh38
rs751335589 Chr.10: 99739684-99739684 on Build GRCh38
rs778689033 Chr.10: 99739795-99739795 on Build GRCh38
rs900787666 Chr.10: 99739780-99739780 on Build GRCh38
rs963767157 Chr.10: 99739630-99739630 on Build GRCh38
rs778111369 Chr.10: 99739688-99739688 on Build GRCh38
rs764577573 Chr.10: 99739766-99739766 on Build GRCh38
rs766534836 Chr.10: 99739662-99739662 on Build GRCh38
rs748835539 Chr.10: 99739701-99739701 on Build GRCh38
rs979481032 Chr.10: 99739636-99739636 on Build GRCh38
rs779620581 Chr.10: 99739695-99739695 on Build GRCh38

View Full Product Details