Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59107485-59107712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
UniGene
Hs.194698
Gene Chromosome Location
Chr.15: 59105085-59125045 on Build GRCh38
Transcripts
NM_004701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622926 c.228T>C p.A76A Chr.15: 59107631-59107631 on Build GRCh38
127568 c.162G>A p.Q54Q Chr.15: 59107565-59107565 on Build GRCh38
5383611 c.210A>G p.K70K Chr.15: 59107613-59107613 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs559386008 Chr.15: 59107546-59107546 on Build GRCh38
rs189971570 Chr.15: 59107554-59107554 on Build GRCh38
rs773159638 Chr.15: 59107569-59107569 on Build GRCh38
rs775731943 Chr.15: 59107517-59107517 on Build GRCh38
rs35033821 Chr.15: 59107670-59107670 on Build GRCh38
rs866249126 Chr.15: 59107588-59107588 on Build GRCh38
rs750754556 Chr.15: 59107537-59107537 on Build GRCh38
rs749059876 Chr.15: 59107571-59107571 on Build GRCh38
rs539408437 Chr.15: 59107552-59107552 on Build GRCh38
rs755571015 Chr.15: 59107679-59107679 on Build GRCh38
rs199782001 Chr.15: 59107631-59107631 on Build GRCh38
rs200529135 Chr.15: 59107611-59107611 on Build GRCh38
rs1021675271 Chr.15: 59107596-59107596 on Build GRCh38
rs34747071 Chr.15: 59107643-59107643 on Build GRCh38
rs921961093 Chr.15: 59107509-59107509 on Build GRCh38
rs144091405 Chr.15: 59107633-59107633 on Build GRCh38
rs767576210 Chr.15: 59107588-59107590 on Build GRCh38
rs146926787 Chr.15: 59107597-59107597 on Build GRCh38
rs369986119 Chr.15: 59107678-59107678 on Build GRCh38
rs138232014 Chr.15: 59107603-59107603 on Build GRCh38
rs765697847 Chr.15: 59107610-59107610 on Build GRCh38
rs779600368 Chr.15: 59107544-59107544 on Build GRCh38
rs772878851 Chr.15: 59107638-59107638 on Build GRCh38
rs775613648 Chr.15: 59107589-59107589 on Build GRCh38
rs761914242 Chr.15: 59107605-59107605 on Build GRCh38
rs760885492 Chr.15: 59107521-59107521 on Build GRCh38
rs138427426 Chr.15: 59107560-59107560 on Build GRCh38
rs201051714 Chr.15: 59107642-59107642 on Build GRCh38
rs757823735 Chr.15: 59107534-59107534 on Build GRCh38
rs369361404 Chr.15: 59107529-59107529 on Build GRCh38
rs1051832905 Chr.15: 59107543-59107543 on Build GRCh38
rs765713696 Chr.15: 59107536-59107536 on Build GRCh38
rs150253971 Chr.15: 59107604-59107604 on Build GRCh38
rs762752818 Chr.15: 59107515-59107515 on Build GRCh38
rs767663609 Chr.15: 59107516-59107516 on Build GRCh38
rs754613732 Chr.15: 59107655-59107655 on Build GRCh38
rs746653521 Chr.15: 59107548-59107548 on Build GRCh38
rs11548100 Chr.15: 59107586-59107586 on Build GRCh38
rs11548098 Chr.15: 59107626-59107626 on Build GRCh38
rs762932691 Chr.15: 59107536-59107536 on Build GRCh38
rs374923143 Chr.15: 59107510-59107510 on Build GRCh38
rs773075874 Chr.15: 59107531-59107531 on Build GRCh38
rs754459555 Chr.15: 59107531-59107531 on Build GRCh38

View Full Product Details