Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40062831-40063086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCOR
Entrez Gene ID
54880
Gene Name
BCL6 corepressor
Gene Aliases
ANOP2, MAA2, MCOPS2
UniGene
Hs.659681
Gene Chromosome Location
Chr.X: 40051246-40177390 on Build GRCh38
Transcripts
NM_001123383.1 NM_001123384.1 NM_001123385.1 NM_017745.5 XM_005272616.1 XM_005272618.3 XM_005272619.3 XM_005272620.3 XM_006724536.3 XM_011543929.2 XM_011543930.1 XM_011543931.2 XM_017029615.1 XM_017029616.1

SNP Detail

SNP ID Chromosome Location
rs768842901 Chr.X: 40062865-40062865 on Build GRCh38
rs981265184 Chr.X: 40062913-40062913 on Build GRCh38
rs747055096 Chr.X: 40062857-40062857 on Build GRCh38
rs780712297 Chr.X: 40062856-40062856 on Build GRCh38
rs187172641 Chr.X: 40062945-40062945 on Build GRCh38
rs760362391 Chr.X: 40062934-40062934 on Build GRCh38
rs142866108 Chr.X: 40062905-40062905 on Build GRCh38
rs749073971 Chr.X: 40063026-40063026 on Build GRCh38
rs972573105 Chr.X: 40062968-40062968 on Build GRCh38
rs763853638 Chr.X: 40062938-40062938 on Build GRCh38
rs886042842 Chr.X: 40063035-40063036 on Build GRCh38
rs905710587 Chr.X: 40063038-40063038 on Build GRCh38
rs370685925 Chr.X: 40062910-40062910 on Build GRCh38
rs748201753 Chr.X: 40062992-40062992 on Build GRCh38
rs777973496 Chr.X: 40063012-40063012 on Build GRCh38
rs868100355 Chr.X: 40063011-40063011 on Build GRCh38
rs930630046 Chr.X: 40062982-40062982 on Build GRCh38
rs772939285 Chr.X: 40062896-40062896 on Build GRCh38
rs771933828 Chr.X: 40063054-40063054 on Build GRCh38
rs767103127 Chr.X: 40062915-40062915 on Build GRCh38
rs199795707 Chr.X: 40063055-40063055 on Build GRCh38
rs770038057 Chr.X: 40062878-40062878 on Build GRCh38
rs781293725 Chr.X: 40062986-40062986 on Build GRCh38
rs908375616 Chr.X: 40062941-40062941 on Build GRCh38
rs752258567 Chr.X: 40062932-40062932 on Build GRCh38
rs752160903 Chr.X: 40062971-40062971 on Build GRCh38
rs758900726 Chr.X: 40062963-40062963 on Build GRCh38
rs781252900 Chr.X: 40062866-40062866 on Build GRCh38
rs373762694 Chr.X: 40062911-40062911 on Build GRCh38
rs371205644 Chr.X: 40063047-40063047 on Build GRCh38
rs888552486 Chr.X: 40062985-40062985 on Build GRCh38
rs748445735 Chr.X: 40062868-40062868 on Build GRCh38
rs754985350 Chr.X: 40062978-40062978 on Build GRCh38
rs774132405 Chr.X: 40063046-40063046 on Build GRCh38
rs372510528 Chr.X: 40062966-40062966 on Build GRCh38
rs770766590 Chr.X: 40063042-40063042 on Build GRCh38
rs376614903 Chr.X: 40062861-40062861 on Build GRCh38
rs769942209 Chr.X: 40063000-40063000 on Build GRCh38
rs780412607 Chr.X: 40062969-40062969 on Build GRCh38
rs770784599 Chr.X: 40062906-40062906 on Build GRCh38

View Full Product Details