Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172769676-172769901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP1
Entrez Gene ID
1843
Gene Name
dual specificity phosphatase 1
Gene Aliases
CL100, HVH1, MKP-1, MKP1, PTPN10
UniGene
Hs.171695
Gene Chromosome Location
Chr.5: 172768090-172771200 on Build GRCh38
Transcripts
NM_004417.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6993336 c.606C>T p.A202A Chr.5: 172769702-172769702 on Build GRCh38
6836661 c.532C>G p.L178V Chr.5: 172769776-172769776 on Build GRCh38
3853747 c.547C>T p.L183L Chr.5: 172769761-172769761 on Build GRCh38
3827694 c.595G>A p.G199S Chr.5: 172769713-172769713 on Build GRCh38
6364007 c.550G>T p.G184C Chr.5: 172769758-172769758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978226470 Chr.5: 172769699-172769699 on Build GRCh38
rs538300481 Chr.5: 172769804-172769804 on Build GRCh38
rs748817760 Chr.5: 172769787-172769787 on Build GRCh38
rs938404789 Chr.5: 172769731-172769731 on Build GRCh38
rs762145525 Chr.5: 172769737-172769737 on Build GRCh38
rs755742331 Chr.5: 172769780-172769780 on Build GRCh38
rs954176133 Chr.5: 172769850-172769850 on Build GRCh38
rs2431663 Chr.5: 172769708-172769708 on Build GRCh38
rs767901692 Chr.5: 172769751-172769751 on Build GRCh38
rs776950475 Chr.5: 172769736-172769736 on Build GRCh38
rs185525501 Chr.5: 172769805-172769805 on Build GRCh38
rs574252357 Chr.5: 172769851-172769851 on Build GRCh38
rs778841658 Chr.5: 172769792-172769792 on Build GRCh38
rs755306894 Chr.5: 172769750-172769750 on Build GRCh38
rs34471628 Chr.5: 172769749-172769749 on Build GRCh38
rs200791815 Chr.5: 172769811-172769811 on Build GRCh38
rs759315876 Chr.5: 172769718-172769718 on Build GRCh38
rs140793690 Chr.5: 172769847-172769847 on Build GRCh38
rs773391773 Chr.5: 172769739-172769739 on Build GRCh38
rs372702653 Chr.5: 172769719-172769719 on Build GRCh38
rs896619361 Chr.5: 172769854-172769854 on Build GRCh38
rs137949520 Chr.5: 172769789-172769789 on Build GRCh38
rs57501400 Chr.5: 172769812-172769812 on Build GRCh38
rs745822861 Chr.5: 172769795-172769795 on Build GRCh38
rs770896245 Chr.5: 172769703-172769703 on Build GRCh38
rs556217277 Chr.5: 172769786-172769786 on Build GRCh38
rs752300180 Chr.5: 172769773-172769773 on Build GRCh38
rs763013896 Chr.5: 172769741-172769741 on Build GRCh38
rs1057281085 Chr.5: 172769734-172769734 on Build GRCh38

View Full Product Details