Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43507871-43508128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETHE1
Entrez Gene ID
23474
Gene Name
ETHE1, persulfide dioxygenase
Gene Aliases
HSCO, YF13H12
UniGene
Hs.7486
Gene Chromosome Location
Chr.19: 43506719-43527256 on Build GRCh38
Transcripts
NM_001320867.1 NM_001320868.1 NM_001320869.1 NM_014297.4 XM_005258687.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390001 c.611C>T p.T204I Chr.19: 43508045-43508045 on Build GRCh38
7166352 c.631C>G p.L211V Chr.19: 43508025-43508025 on Build GRCh38
4648288 c.697A>G p.K233E Chr.19: 43507959-43507959 on Build GRCh38
5921423 c.682A>C p.N228H Chr.19: 43507974-43507974 on Build GRCh38
1750954 c.596G>C p.G199A Chr.19: 43508060-43508060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042019932 Chr.19: 43507956-43507956 on Build GRCh38
rs778245630 Chr.19: 43507978-43507978 on Build GRCh38
rs749639950 Chr.19: 43507994-43507994 on Build GRCh38
rs775667650 Chr.19: 43508035-43508035 on Build GRCh38
rs369625358 Chr.19: 43507972-43507972 on Build GRCh38
rs113608853 Chr.19: 43507892-43507892 on Build GRCh38
rs758313443 Chr.19: 43507930-43507930 on Build GRCh38
rs779462872 Chr.19: 43508101-43508101 on Build GRCh38
rs766269215 Chr.19: 43508054-43508054 on Build GRCh38
rs745956051 Chr.19: 43508026-43508026 on Build GRCh38
rs754448418 Chr.19: 43507949-43507949 on Build GRCh38
rs374400582 Chr.19: 43508096-43508096 on Build GRCh38
rs762667429 Chr.19: 43508053-43508053 on Build GRCh38
rs751515256 Chr.19: 43508067-43508067 on Build GRCh38
rs200319834 Chr.19: 43507898-43507898 on Build GRCh38
rs901230825 Chr.19: 43508059-43508059 on Build GRCh38
rs1054773943 Chr.19: 43507920-43507920 on Build GRCh38
rs267605525 Chr.19: 43508048-43508048 on Build GRCh38
rs771413307 Chr.19: 43508005-43508005 on Build GRCh38
rs863223954 Chr.19: 43508062-43508062 on Build GRCh38
rs774726187 Chr.19: 43508019-43508019 on Build GRCh38
rs201485073 Chr.19: 43508097-43508097 on Build GRCh38
rs1023744474 Chr.19: 43508102-43508102 on Build GRCh38
rs902352738 Chr.19: 43508027-43508027 on Build GRCh38
rs1050192297 Chr.19: 43507909-43507909 on Build GRCh38
rs939026601 Chr.19: 43507915-43507915 on Build GRCh38
rs779584627 Chr.19: 43508015-43508015 on Build GRCh38
rs766247547 Chr.19: 43507941-43507941 on Build GRCh38
rs756520191 Chr.19: 43507974-43507974 on Build GRCh38
rs863223956 Chr.19: 43508053-43508054 on Build GRCh38
rs200179864 Chr.19: 43508099-43508099 on Build GRCh38
rs182058313 Chr.19: 43507961-43507961 on Build GRCh38
rs952297351 Chr.19: 43508106-43508106 on Build GRCh38
rs202162263 Chr.19: 43508091-43508091 on Build GRCh38
rs751021285 Chr.19: 43507945-43507945 on Build GRCh38
rs757630152 Chr.19: 43508100-43508100 on Build GRCh38
rs373746054 Chr.19: 43508044-43508044 on Build GRCh38
rs765064380 Chr.19: 43507908-43507908 on Build GRCh38
rs972873758 Chr.19: 43507895-43507895 on Build GRCh38
rs759099758 Chr.19: 43508068-43508068 on Build GRCh38
rs946602889 Chr.19: 43507963-43507963 on Build GRCh38
rs772197337 Chr.19: 43508029-43508029 on Build GRCh38
rs1056355030 Chr.19: 43508047-43508047 on Build GRCh38
rs1002746898 Chr.19: 43508018-43508018 on Build GRCh38
rs995933852 Chr.19: 43507985-43507985 on Build GRCh38
rs767180237 Chr.19: 43508085-43508085 on Build GRCh38
rs773009938 Chr.19: 43508051-43508051 on Build GRCh38
rs375984717 Chr.19: 43507911-43507911 on Build GRCh38

View Full Product Details