Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152720226-152720469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf68
Entrez Gene ID
100129271
Gene Name
chromosome 1 open reading frame 68
Gene Aliases
LEP7, XP32
UniGene
Hs.601077
Gene Chromosome Location
Chr.1: 152719522-152720429 on Build GRCh38
Transcripts
NM_001024679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5377685 c.729C>T p.D243D Chr.1: 152720250-152720250 on Build GRCh38
3705001 c.744C>T p.C248C Chr.1: 152720265-152720265 on Build GRCh38
4998099 c.716C>G p.P239R Chr.1: 152720237-152720237 on Build GRCh38
2207263 c.739G>A p.D247N Chr.1: 152720260-152720260 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370263286 Chr.1: 152720336-152720336 on Build GRCh38
rs3814351 Chr.1: 152720386-152720386 on Build GRCh38
rs957306846 Chr.1: 152720374-152720374 on Build GRCh38
rs774508921 Chr.1: 152720299-152720299 on Build GRCh38
rs773547274 Chr.1: 152720280-152720280 on Build GRCh38
rs749560310 Chr.1: 152720281-152720281 on Build GRCh38
rs781353683 Chr.1: 152720244-152720244 on Build GRCh38
rs1026865002 Chr.1: 152720302-152720302 on Build GRCh38
rs919772786 Chr.1: 152720295-152720295 on Build GRCh38
rs893302288 Chr.1: 152720441-152720441 on Build GRCh38
rs751331955 Chr.1: 152720309-152720309 on Build GRCh38
rs183266025 Chr.1: 152720289-152720289 on Build GRCh38
rs543308081 Chr.1: 152720421-152720421 on Build GRCh38
rs3814350 Chr.1: 152720305-152720305 on Build GRCh38
rs41268476 Chr.1: 152720327-152720327 on Build GRCh38
rs748518993 Chr.1: 152720258-152720258 on Build GRCh38
rs964363047 Chr.1: 152720334-152720334 on Build GRCh38
rs913025490 Chr.1: 152720405-152720405 on Build GRCh38
rs745402357 Chr.1: 152720368-152720368 on Build GRCh38
rs770340863 Chr.1: 152720260-152720260 on Build GRCh38
rs776620137 Chr.1: 152720317-152720317 on Build GRCh38

View Full Product Details