Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10118384-10118657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC7A
Entrez Gene ID
64581
Gene Name
C-type lectin domain family 7 member A
Gene Aliases
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2
UniGene
Hs.143929
Gene Chromosome Location
Chr.12: 10116777-10130269 on Build GRCh38
Transcripts
NM_022570.4 NM_197947.2 NM_197948.2 NM_197949.2 NM_197950.2 NR_125336.1 XM_006719135.3 XM_017019822.1 XM_017019823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6477614 c.535G>T p.V179L Chr.12: 10118529-10118529 on Build GRCh38
5952037 c.740T>C p.M247T Chr.12: 10118462-10118462 on Build GRCh38
6630253 c.537G>A p.V179V Chr.12: 10118527-10118527 on Build GRCh38
3954064 c.590A>T p.K197M Chr.12: 10118474-10118474 on Build GRCh38
6630251 c.534C>T p.H178H Chr.12: 10118530-10118530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761669631 Chr.12: 10118529-10118529 on Build GRCh38
rs770612544 Chr.12: 10118423-10118423 on Build GRCh38
rs750471406 Chr.12: 10118471-10118471 on Build GRCh38
rs776324378 Chr.12: 10118470-10118472 on Build GRCh38
rs760732103 Chr.12: 10118537-10118537 on Build GRCh38
rs763991603 Chr.12: 10118449-10118449 on Build GRCh38
rs112789481 Chr.12: 10118524-10118524 on Build GRCh38
rs531718245 Chr.12: 10118566-10118566 on Build GRCh38
rs201458154 Chr.12: 10118619-10118619 on Build GRCh38
rs151164069 Chr.12: 10118463-10118463 on Build GRCh38
rs766564798 Chr.12: 10118486-10118486 on Build GRCh38
rs780697158 Chr.12: 10118495-10118495 on Build GRCh38
rs769484555 Chr.12: 10118507-10118507 on Build GRCh38
rs7959451 Chr.12: 10118456-10118456 on Build GRCh38
rs771913423 Chr.12: 10118530-10118530 on Build GRCh38
rs374910100 Chr.12: 10118557-10118557 on Build GRCh38
rs747946367 Chr.12: 10118634-10118634 on Build GRCh38
rs749063912 Chr.12: 10118522-10118522 on Build GRCh38
rs758834241 Chr.12: 10118492-10118492 on Build GRCh38
rs202170690 Chr.12: 10118612-10118612 on Build GRCh38
rs16910527 Chr.12: 10118534-10118534 on Build GRCh38
rs745459200 Chr.12: 10118430-10118430 on Build GRCh38
rs753128502 Chr.12: 10118575-10118575 on Build GRCh38
rs748902268 Chr.12: 10118618-10118618 on Build GRCh38
rs1022483302 Chr.12: 10118582-10118582 on Build GRCh38
rs754160847 Chr.12: 10118545-10118545 on Build GRCh38
rs769346152 Chr.12: 10118436-10118436 on Build GRCh38
rs755633133 Chr.12: 10118607-10118607 on Build GRCh38
rs749921680 Chr.12: 10118602-10118602 on Build GRCh38
rs760890056 Chr.12: 10118485-10118485 on Build GRCh38
rs778211636 Chr.12: 10118590-10118590 on Build GRCh38
rs16910526 Chr.12: 10118488-10118488 on Build GRCh38
rs775343205 Chr.12: 10118437-10118437 on Build GRCh38
rs762806010 Chr.12: 10118446-10118446 on Build GRCh38
rs982339053 Chr.12: 10118448-10118448 on Build GRCh38
rs746485235 Chr.12: 10118419-10118419 on Build GRCh38
rs893595643 Chr.12: 10118604-10118604 on Build GRCh38
rs758931112 Chr.12: 10118589-10118589 on Build GRCh38
rs776038688 Chr.12: 10118428-10118428 on Build GRCh38
rs543022264 Chr.12: 10118528-10118528 on Build GRCh38
rs371994700 Chr.12: 10118538-10118538 on Build GRCh38
rs753204580 Chr.12: 10118491-10118491 on Build GRCh38
rs141153031 Chr.12: 10118504-10118504 on Build GRCh38
rs779538254 Chr.12: 10118490-10118490 on Build GRCh38
rs144851220 Chr.12: 10118526-10118526 on Build GRCh38
rs916828362 Chr.12: 10118417-10118417 on Build GRCh38
rs779616817 Chr.12: 10118509-10118509 on Build GRCh38
rs999231169 Chr.12: 10118583-10118583 on Build GRCh38
rs778619699 Chr.12: 10118630-10118630 on Build GRCh38

View Full Product Details