Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43789518-43789791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
UniGene
Hs.371597
Gene Chromosome Location
Chr.2: 43774039-43828492 on Build GRCh38
Transcripts
NM_001193464.1 NM_015522.3 NM_016008.3 XM_005264364.4 XM_005264365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5354742 c.284A>G p.D95G Chr.2: 43789685-43789685 on Build GRCh38
4480657 c.243C>T p.I81I Chr.2: 43789644-43789644 on Build GRCh38
1690524 c.252T>A p.F84L Chr.2: 43789653-43789653 on Build GRCh38
5677292 c.306A>G p.T102T Chr.2: 43789707-43789707 on Build GRCh38
6091502 c.283G>T p.D95Y Chr.2: 43789684-43789684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759829586 Chr.2: 43789753-43789753 on Build GRCh38
rs370305575 Chr.2: 43789760-43789760 on Build GRCh38
rs745454160 Chr.2: 43789617-43789617 on Build GRCh38
rs140794515 Chr.2: 43789586-43789586 on Build GRCh38
rs765666851 Chr.2: 43789670-43789670 on Build GRCh38
rs201121259 Chr.2: 43789662-43789662 on Build GRCh38
rs763433820 Chr.2: 43789757-43789757 on Build GRCh38
rs3815994 Chr.2: 43789746-43789746 on Build GRCh38
rs748999743 Chr.2: 43789711-43789711 on Build GRCh38
rs753156802 Chr.2: 43789673-43789673 on Build GRCh38
rs753521543 Chr.2: 43789647-43789647 on Build GRCh38
rs749989492 Chr.2: 43789687-43789687 on Build GRCh38
rs748116445 Chr.2: 43789640-43789640 on Build GRCh38
rs879025579 Chr.2: 43789747-43789747 on Build GRCh38
rs772219395 Chr.2: 43789641-43789641 on Build GRCh38
rs776679741 Chr.2: 43789749-43789749 on Build GRCh38
rs767007849 Chr.2: 43789686-43789686 on Build GRCh38
rs559156619 Chr.2: 43789666-43789666 on Build GRCh38
rs755765538 Chr.2: 43789696-43789696 on Build GRCh38
rs986143133 Chr.2: 43789599-43789600 on Build GRCh38
rs374265551 Chr.2: 43789663-43789663 on Build GRCh38
rs559666542 Chr.2: 43789601-43789601 on Build GRCh38
rs184689483 Chr.2: 43789762-43789762 on Build GRCh38
rs370458576 Chr.2: 43789603-43789603 on Build GRCh38
rs751042370 Chr.2: 43789611-43789611 on Build GRCh38
rs779882438 Chr.2: 43789630-43789630 on Build GRCh38
rs754679765 Chr.2: 43789713-43789713 on Build GRCh38
rs753395636 Chr.2: 43789684-43789684 on Build GRCh38
rs376082024 Chr.2: 43789590-43789590 on Build GRCh38
rs779612644 Chr.2: 43789705-43789705 on Build GRCh38
rs761417614 Chr.2: 43789749-43789749 on Build GRCh38
rs749081820 Chr.2: 43789631-43789631 on Build GRCh38
rs755498989 Chr.2: 43789602-43789602 on Build GRCh38
rs760992508 Chr.2: 43789587-43789587 on Build GRCh38
rs775896619 Chr.2: 43789756-43789756 on Build GRCh38
rs772202929 Chr.2: 43789721-43789721 on Build GRCh38
rs550471562 Chr.2: 43789733-43789733 on Build GRCh38
rs202154744 Chr.2: 43789613-43789613 on Build GRCh38
rs748032561 Chr.2: 43789720-43789720 on Build GRCh38
rs903394706 Chr.2: 43789764-43789764 on Build GRCh38
rs766568925 Chr.2: 43789659-43789659 on Build GRCh38
rs769397371 Chr.2: 43789625-43789625 on Build GRCh38
rs765530877 Chr.2: 43789754-43789754 on Build GRCh38
rs760977013 Chr.2: 43789645-43789645 on Build GRCh38
rs970811681 Chr.2: 43789622-43789622 on Build GRCh38
rs767734280 Chr.2: 43789584-43789584 on Build GRCh38
rs778864477 Chr.2: 43789716-43789716 on Build GRCh38
rs768606087 Chr.2: 43789634-43789634 on Build GRCh38
rs557553902 Chr.2: 43789575-43789575 on Build GRCh38
rs142464485 Chr.2: 43789685-43789685 on Build GRCh38
rs544639319 Chr.2: 43789644-43789644 on Build GRCh38

View Full Product Details