Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48750476-48750743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT1
Entrez Gene ID
2523
Gene Name
fucosyltransferase 1 (H blood group)
Gene Aliases
H, HH, HSC
UniGene
Hs.69747
Gene Chromosome Location
Chr.19: 48747826-48755390 on Build GRCh38
Transcripts
NM_000148.3 XM_006723127.2 XM_017026552.1 XM_017026553.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3716503 c.658C>T p.R220C Chr.19: 48750624-48750624 on Build GRCh38
6667856 c.804T>C p.C268C Chr.19: 48750478-48750478 on Build GRCh38
1751036 c.696G>A p.W232* Chr.19: 48750586-48750586 on Build GRCh38
6667857 c.689A>C p.Q230P Chr.19: 48750593-48750593 on Build GRCh38
3719885 c.616C>T p.R206C Chr.19: 48750666-48750666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756422637 Chr.19: 48750707-48750707 on Build GRCh38
rs749593829 Chr.19: 48750514-48750514 on Build GRCh38
rs753966600 Chr.19: 48750708-48750708 on Build GRCh38
rs760125997 Chr.19: 48750554-48750554 on Build GRCh38
rs111500075 Chr.19: 48750723-48750723 on Build GRCh38
rs753519856 Chr.19: 48750523-48750523 on Build GRCh38
rs915531286 Chr.19: 48750649-48750649 on Build GRCh38
rs267605568 Chr.19: 48750650-48750650 on Build GRCh38
rs779427274 Chr.19: 48750718-48750718 on Build GRCh38
rs757082412 Chr.19: 48750582-48750582 on Build GRCh38
rs759173829 Chr.19: 48750519-48750519 on Build GRCh38
rs750295143 Chr.19: 48750577-48750577 on Build GRCh38
rs781314062 Chr.19: 48750498-48750498 on Build GRCh38
rs770291377 Chr.19: 48750503-48750503 on Build GRCh38
rs752769882 Chr.19: 48750613-48750613 on Build GRCh38
rs182456777 Chr.19: 48750717-48750717 on Build GRCh38
rs761822435 Chr.19: 48750661-48750661 on Build GRCh38
rs574691621 Chr.19: 48750624-48750624 on Build GRCh38
rs989654136 Chr.19: 48750605-48750605 on Build GRCh38
rs541722036 Chr.19: 48750633-48750633 on Build GRCh38
rs556345040 Chr.19: 48750591-48750591 on Build GRCh38
rs765114567 Chr.19: 48750561-48750561 on Build GRCh38
rs752865322 Chr.19: 48750697-48750697 on Build GRCh38
rs748220172 Chr.19: 48750499-48750499 on Build GRCh38
rs771483957 Chr.19: 48750634-48750634 on Build GRCh38
rs897056205 Chr.19: 48750700-48750700 on Build GRCh38
rs558351055 Chr.19: 48750533-48750533 on Build GRCh38
rs376911171 Chr.19: 48750590-48750590 on Build GRCh38
rs779225361 Chr.19: 48750635-48750635 on Build GRCh38
rs750207214 Chr.19: 48750665-48750665 on Build GRCh38
rs756157363 Chr.19: 48750614-48750614 on Build GRCh38
rs577050576 Chr.19: 48750556-48750556 on Build GRCh38
rs762860495 Chr.19: 48750666-48750666 on Build GRCh38
rs765309154 Chr.19: 48750663-48750663 on Build GRCh38
rs572327966 Chr.19: 48750675-48750675 on Build GRCh38
rs1050483060 Chr.19: 48750567-48750567 on Build GRCh38
rs760165583 Chr.19: 48750656-48750656 on Build GRCh38
rs749787906 Chr.19: 48750628-48750628 on Build GRCh38
rs200299477 Chr.19: 48750518-48750518 on Build GRCh38
rs28934588 Chr.19: 48750557-48750557 on Build GRCh38
rs746271520 Chr.19: 48750637-48750637 on Build GRCh38
rs775020226 Chr.19: 48750654-48750654 on Build GRCh38
rs771713990 Chr.19: 48750648-48750648 on Build GRCh38
rs776265416 Chr.19: 48750658-48750658 on Build GRCh38
rs775034079 Chr.19: 48750545-48750545 on Build GRCh38
rs778160774 Chr.19: 48750507-48750507 on Build GRCh38
rs199502509 Chr.19: 48750657-48750657 on Build GRCh38

View Full Product Details