Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45872351-45872588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA3
Entrez Gene ID
3171
Gene Name
forkhead box A3
Gene Aliases
FKHH3, HNF3G, TCF3G
UniGene
Hs.36137
Gene Chromosome Location
Chr.19: 45864260-45873797 on Build GRCh38
Transcripts
NM_004497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
248772 c.512T>C p.F171S Chr.19: 45872517-45872517 on Build GRCh38
5800770 c.518A>G p.D173G Chr.19: 45872523-45872523 on Build GRCh38
4132148 c.516C>T p.N172N Chr.19: 45872521-45872521 on Build GRCh38
5856196 c.386C>T p.A129V Chr.19: 45872391-45872391 on Build GRCh38
7179801 c.362C>G p.S121C Chr.19: 45872367-45872367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759167859 Chr.19: 45872556-45872556 on Build GRCh38
rs764274524 Chr.19: 45872388-45872388 on Build GRCh38
rs141446079 Chr.19: 45872498-45872498 on Build GRCh38
rs974481837 Chr.19: 45872387-45872387 on Build GRCh38
rs199787706 Chr.19: 45872497-45872497 on Build GRCh38
rs753914826 Chr.19: 45872403-45872403 on Build GRCh38
rs12977291 Chr.19: 45872488-45872488 on Build GRCh38
rs762026809 Chr.19: 45872526-45872526 on Build GRCh38
rs748746722 Chr.19: 45872461-45872461 on Build GRCh38
rs751569343 Chr.19: 45872531-45872531 on Build GRCh38
rs772555492 Chr.19: 45872483-45872483 on Build GRCh38
rs141782286 Chr.19: 45872404-45872404 on Build GRCh38
rs767496597 Chr.19: 45872511-45872511 on Build GRCh38
rs926920586 Chr.19: 45872475-45872475 on Build GRCh38
rs12976619 Chr.19: 45872492-45872492 on Build GRCh38
rs780833457 Chr.19: 45872452-45872452 on Build GRCh38
rs973562577 Chr.19: 45872515-45872515 on Build GRCh38
rs775650612 Chr.19: 45872484-45872484 on Build GRCh38
rs373998257 Chr.19: 45872542-45872542 on Build GRCh38
rs372030164 Chr.19: 45872566-45872566 on Build GRCh38
rs755785237 Chr.19: 45872551-45872551 on Build GRCh38
rs771457315 Chr.19: 45872468-45872468 on Build GRCh38
rs772303969 Chr.19: 45872371-45872371 on Build GRCh38
rs777278399 Chr.19: 45872458-45872458 on Build GRCh38
rs751423514 Chr.19: 45872439-45872439 on Build GRCh38
rs867687500 Chr.19: 45872501-45872501 on Build GRCh38
rs16980091 Chr.19: 45872521-45872521 on Build GRCh38
rs765382731 Chr.19: 45872530-45872530 on Build GRCh38
rs954700998 Chr.19: 45872400-45872400 on Build GRCh38
rs946035893 Chr.19: 45872554-45872554 on Build GRCh38
rs746349583 Chr.19: 45872471-45872471 on Build GRCh38
rs766381222 Chr.19: 45872407-45872407 on Build GRCh38
rs865905914 Chr.19: 45872395-45872395 on Build GRCh38
rs760803594 Chr.19: 45872381-45872381 on Build GRCh38
rs755697325 Chr.19: 45872453-45872453 on Build GRCh38
rs775840325 Chr.19: 45872378-45872378 on Build GRCh38
rs560929375 Chr.19: 45872430-45872430 on Build GRCh38
rs752376903 Chr.19: 45872544-45872544 on Build GRCh38
rs200377130 Chr.19: 45872469-45872469 on Build GRCh38
rs1044654245 Chr.19: 45872448-45872448 on Build GRCh38
rs865961018 Chr.19: 45872552-45872552 on Build GRCh38
rs374769245 Chr.19: 45872446-45872446 on Build GRCh38

View Full Product Details