Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27517913-27518186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX2
Entrez Gene ID
2053
Gene Name
epoxide hydrolase 2
Gene Aliases
CEH, SEH
UniGene
Hs.212088
Gene Chromosome Location
Chr.8: 27491002-27597559 on Build GRCh38
Transcripts
NM_001256482.1 NM_001256483.1 NM_001256484.1 NM_001979.5 XM_017013199.1

SNP Detail

SNP ID Chromosome Location
rs188474787 Chr.8: 27517969-27517969 on Build GRCh38
rs72475851 Chr.8: 27517946-27517946 on Build GRCh38
rs575234938 Chr.8: 27518075-27518075 on Build GRCh38
rs1010347239 Chr.8: 27518158-27518158 on Build GRCh38
rs899978770 Chr.8: 27518099-27518099 on Build GRCh38
rs527622076 Chr.8: 27518163-27518163 on Build GRCh38
rs377587775 Chr.8: 27517961-27517961 on Build GRCh38
rs370696658 Chr.8: 27518013-27518013 on Build GRCh38
rs892711872 Chr.8: 27518140-27518140 on Build GRCh38
rs1008920382 Chr.8: 27518162-27518162 on Build GRCh38
rs373619581 Chr.8: 27518090-27518090 on Build GRCh38
rs755731146 Chr.8: 27518120-27518120 on Build GRCh38
rs72475852 Chr.8: 27518109-27518109 on Build GRCh38
rs927018633 Chr.8: 27517973-27517973 on Build GRCh38
rs929999026 Chr.8: 27517977-27517977 on Build GRCh38
rs377527949 Chr.8: 27518002-27518002 on Build GRCh38
rs769013171 Chr.8: 27518049-27518052 on Build GRCh38
rs561646330 Chr.8: 27518131-27518131 on Build GRCh38
rs758291257 Chr.8: 27518080-27518080 on Build GRCh38
rs146084287 Chr.8: 27518067-27518067 on Build GRCh38
rs2234914 Chr.8: 27518048-27518048 on Build GRCh38
rs201843582 Chr.8: 27518110-27518110 on Build GRCh38
rs754451478 Chr.8: 27518094-27518094 on Build GRCh38
rs750223134 Chr.8: 27518077-27518077 on Build GRCh38
rs1036564154 Chr.8: 27518073-27518073 on Build GRCh38
rs766324746 Chr.8: 27518089-27518089 on Build GRCh38
rs775752053 Chr.8: 27518018-27518018 on Build GRCh38
rs749860220 Chr.8: 27518046-27518047 on Build GRCh38
rs778136742 Chr.8: 27518121-27518121 on Build GRCh38
rs374441457 Chr.8: 27518130-27518130 on Build GRCh38
rs896072745 Chr.8: 27518046-27518046 on Build GRCh38
rs759713879 Chr.8: 27518011-27518011 on Build GRCh38
rs572825643 Chr.8: 27517942-27517942 on Build GRCh38
rs796444961 Chr.8: 27518042-27518042 on Build GRCh38
rs145198899 Chr.8: 27517967-27517967 on Build GRCh38

View Full Product Details