Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113369706-113369951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSPRY
Entrez Gene ID
54836
Gene Name
B-box and SPRY domain containing
Gene Aliases
-
UniGene
Hs.614517
Gene Chromosome Location
Chr.9: 113349532-113371233 on Build GRCh38
Transcripts
NM_001317943.1 NM_001317944.1 NM_017688.2 XM_006717149.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3395510 c.783C>A p.A261A Chr.9: 113369716-113369716 on Build GRCh38
7447726 c.795delC p.P266fs*25 Chr.9: 113369728-113369728 on Build GRCh38
7357804 c.781G>C p.A261P Chr.9: 113369714-113369714 on Build GRCh38
4666305 c.949C>T p.R317C Chr.9: 113369882-113369882 on Build GRCh38
6182022 c.795C>T p.G265G Chr.9: 113369728-113369728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749579241 Chr.9: 113369833-113369833 on Build GRCh38
rs192218285 Chr.9: 113369743-113369743 on Build GRCh38
rs371439111 Chr.9: 113369817-113369817 on Build GRCh38
rs776900968 Chr.9: 113369847-113369847 on Build GRCh38
rs765270513 Chr.9: 113369803-113369803 on Build GRCh38
rs552798374 Chr.9: 113369774-113369774 on Build GRCh38
rs762277492 Chr.9: 113369800-113369800 on Build GRCh38
rs201947061 Chr.9: 113369728-113369728 on Build GRCh38
rs772538418 Chr.9: 113369725-113369725 on Build GRCh38
rs818711 Chr.9: 113369812-113369812 on Build GRCh38
rs781182313 Chr.9: 113369753-113369753 on Build GRCh38
rs747334548 Chr.9: 113369790-113369790 on Build GRCh38
rs756835090 Chr.9: 113369837-113369837 on Build GRCh38
rs780241775 Chr.9: 113369738-113369738 on Build GRCh38
rs763033041 Chr.9: 113369809-113369809 on Build GRCh38
rs754303460 Chr.9: 113369734-113369734 on Build GRCh38
rs202088728 Chr.9: 113369918-113369918 on Build GRCh38
rs200321662 Chr.9: 113369854-113369854 on Build GRCh38
rs774163931 Chr.9: 113369782-113369782 on Build GRCh38
rs371056236 Chr.9: 113369792-113369792 on Build GRCh38
rs559226374 Chr.9: 113369836-113369836 on Build GRCh38
rs756071553 Chr.9: 113369820-113369820 on Build GRCh38
rs755989896 Chr.9: 113369927-113369927 on Build GRCh38
rs748843741 Chr.9: 113369765-113369765 on Build GRCh38
rs766501173 Chr.9: 113369887-113369887 on Build GRCh38
rs778625266 Chr.9: 113369838-113369838 on Build GRCh38
rs765358310 Chr.9: 113369736-113369736 on Build GRCh38
rs373152755 Chr.9: 113369855-113369855 on Build GRCh38
rs774344945 Chr.9: 113369923-113369929 on Build GRCh38
rs768751053 Chr.9: 113369913-113369914 on Build GRCh38
rs375409553 Chr.9: 113369744-113369744 on Build GRCh38
rs777880341 Chr.9: 113369833-113369833 on Build GRCh38
rs748091270 Chr.9: 113369754-113369754 on Build GRCh38
rs971699028 Chr.9: 113369858-113369858 on Build GRCh38
rs763014503 Chr.9: 113369883-113369883 on Build GRCh38
rs773924146 Chr.9: 113369859-113369859 on Build GRCh38
rs368406613 Chr.9: 113369768-113369768 on Build GRCh38
rs750621245 Chr.9: 113369740-113369740 on Build GRCh38
rs983327955 Chr.9: 113369784-113369784 on Build GRCh38
rs533038118 Chr.9: 113369882-113369882 on Build GRCh38
rs777183976 Chr.9: 113369797-113369797 on Build GRCh38
rs773227381 Chr.9: 113369807-113369807 on Build GRCh38
rs752645388 Chr.9: 113369819-113369819 on Build GRCh38
rs367978873 Chr.9: 113369815-113369815 on Build GRCh38
rs769043503 Chr.9: 113369844-113369844 on Build GRCh38
rs372248622 Chr.9: 113369919-113369919 on Build GRCh38
rs769402253 Chr.9: 113369757-113369757 on Build GRCh38
rs201165135 Chr.9: 113369732-113369732 on Build GRCh38
rs921678137 Chr.9: 113369804-113369804 on Build GRCh38
rs758660546 Chr.9: 113369741-113369741 on Build GRCh38
rs745567321 Chr.9: 113369840-113369840 on Build GRCh38
rs760966749 Chr.9: 113369731-113369731 on Build GRCh38
rs374732028 Chr.9: 113369818-113369818 on Build GRCh38
rs774607143 Chr.9: 113369895-113369895 on Build GRCh38
rs751793885 Chr.9: 113369816-113369816 on Build GRCh38
rs973479711 Chr.9: 113369802-113369802 on Build GRCh38
rs759714752 Chr.9: 113369907-113369907 on Build GRCh38
rs371273259 Chr.9: 113369730-113369730 on Build GRCh38
rs948571051 Chr.9: 113369924-113369924 on Build GRCh38
rs201724584 Chr.9: 113369735-113369735 on Build GRCh38
rs770228595 Chr.9: 113369857-113369857 on Build GRCh38

View Full Product Details