Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1478877-1479112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf25
Entrez Gene ID
148223
Gene Name
chromosome 19 open reading frame 25
Gene Aliases
-
UniGene
Hs.532840
Gene Chromosome Location
Chr.19: 1473201-1479552 on Build GRCh38
Transcripts
NM_152482.2 XM_005259506.3 XM_006722653.3 XM_017026374.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381621 c.21G>T p.K7N Chr.19: 1478883-1478883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950981170 Chr.19: 1478983-1478983 on Build GRCh38
rs761314682 Chr.19: 1478947-1478947 on Build GRCh38
rs768115945 Chr.19: 1478940-1478940 on Build GRCh38
rs921680341 Chr.19: 1479058-1479058 on Build GRCh38
rs778049476 Chr.19: 1478920-1478920 on Build GRCh38
rs1039587888 Chr.19: 1479020-1479020 on Build GRCh38
rs774884778 Chr.19: 1478934-1478934 on Build GRCh38
rs573878652 Chr.19: 1478924-1478924 on Build GRCh38
rs370991961 Chr.19: 1478949-1478949 on Build GRCh38
rs984036850 Chr.19: 1478987-1478987 on Build GRCh38
rs944455712 Chr.19: 1479002-1479002 on Build GRCh38
rs147100215 Chr.19: 1478922-1478922 on Build GRCh38
rs569655428 Chr.19: 1478898-1478898 on Build GRCh38
rs572164899 Chr.19: 1479089-1479089 on Build GRCh38
rs534504395 Chr.19: 1478973-1478973 on Build GRCh38
rs756346092 Chr.19: 1478916-1478916 on Build GRCh38
rs781475466 Chr.19: 1478914-1478914 on Build GRCh38
rs751638266 Chr.19: 1478895-1478895 on Build GRCh38
rs999596890 Chr.19: 1478966-1478966 on Build GRCh38
rs746493920 Chr.19: 1478939-1478939 on Build GRCh38
rs935709973 Chr.19: 1479087-1479087 on Build GRCh38
rs1027931987 Chr.19: 1478960-1478960 on Build GRCh38
rs536913103 Chr.19: 1478911-1478911 on Build GRCh38
rs912038968 Chr.19: 1478995-1478995 on Build GRCh38
rs1003607985 Chr.19: 1478900-1478900 on Build GRCh38
rs974432171 Chr.19: 1479056-1479056 on Build GRCh38
rs552787327 Chr.19: 1479071-1479071 on Build GRCh38
rs776332475 Chr.19: 1478946-1478946 on Build GRCh38

View Full Product Details