Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27744609-27744852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM76A
Entrez Gene ID
199870
Gene Name
family with sequence similarity 76 member A
Gene Aliases
-
UniGene
Hs.469359
Gene Chromosome Location
Chr.1: 27725979-27762915 on Build GRCh38
Transcripts
NM_001143912.1 NM_001143913.1 NM_001143914.1 NM_001143915.1 NM_152660.2 XM_017000543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751729 c.380T>G p.L127R Chr.1: 27744679-27744679 on Build GRCh38
4236357 c.521A>G p.K174R Chr.1: 27744718-27744718 on Build GRCh38
2236392 c.419A>G p.K140R Chr.1: 27744718-27744718 on Build GRCh38
3487724 c.484C>T p.R162C Chr.1: 27744783-27744783 on Build GRCh38
7090570 c.451C>A p.R151S Chr.1: 27744750-27744750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772822320 Chr.1: 27744807-27744807 on Build GRCh38
rs776749545 Chr.1: 27744686-27744686 on Build GRCh38
rs766190202 Chr.1: 27744709-27744709 on Build GRCh38
rs1007517413 Chr.1: 27744699-27744699 on Build GRCh38
rs748057085 Chr.1: 27744784-27744784 on Build GRCh38
rs769661983 Chr.1: 27744697-27744697 on Build GRCh38
rs1022632122 Chr.1: 27744799-27744799 on Build GRCh38
rs771217214 Chr.1: 27744671-27744671 on Build GRCh38
rs113490357 Chr.1: 27744738-27744738 on Build GRCh38
rs970712791 Chr.1: 27744820-27744820 on Build GRCh38
rs778036452 Chr.1: 27744656-27744656 on Build GRCh38
rs999976143 Chr.1: 27744684-27744684 on Build GRCh38
rs778742690 Chr.1: 27744750-27744750 on Build GRCh38
rs184211917 Chr.1: 27744680-27744680 on Build GRCh38
rs904331891 Chr.1: 27744645-27744645 on Build GRCh38
rs766066251 Chr.1: 27744715-27744715 on Build GRCh38
rs146097753 Chr.1: 27744774-27744774 on Build GRCh38
rs898605567 Chr.1: 27744653-27744653 on Build GRCh38
rs572625727 Chr.1: 27744661-27744661 on Build GRCh38
rs766408473 Chr.1: 27744642-27744644 on Build GRCh38
rs754235619 Chr.1: 27744744-27744744 on Build GRCh38
rs747937419 Chr.1: 27744751-27744751 on Build GRCh38
rs987822654 Chr.1: 27744764-27744764 on Build GRCh38
rs762443177 Chr.1: 27744704-27744704 on Build GRCh38
rs748511825 Chr.1: 27744641-27744641 on Build GRCh38
rs780303546 Chr.1: 27744782-27744782 on Build GRCh38
rs754748633 Chr.1: 27744745-27744745 on Build GRCh38
rs756638402 Chr.1: 27744734-27744734 on Build GRCh38
rs200030203 Chr.1: 27744827-27744827 on Build GRCh38
rs375940293 Chr.1: 27744790-27744790 on Build GRCh38
rs753817236 Chr.1: 27744728-27744728 on Build GRCh38
rs767386979 Chr.1: 27744727-27744727 on Build GRCh38
rs931286706 Chr.1: 27744666-27744666 on Build GRCh38
rs777962024 Chr.1: 27744737-27744737 on Build GRCh38
rs761679066 Chr.1: 27744692-27744692 on Build GRCh38
rs533452379 Chr.1: 27744719-27744719 on Build GRCh38
rs745974264 Chr.1: 27744674-27744674 on Build GRCh38
rs773089779 Chr.1: 27744700-27744700 on Build GRCh38
rs374146276 Chr.1: 27744828-27744828 on Build GRCh38
rs981746479 Chr.1: 27744821-27744821 on Build GRCh38
rs751662177 Chr.1: 27744782-27744783 on Build GRCh38
rs1049726533 Chr.1: 27744676-27744676 on Build GRCh38
rs192347522 Chr.1: 27744771-27744771 on Build GRCh38
rs189399027 Chr.1: 27744718-27744718 on Build GRCh38
rs756515578 Chr.1: 27744646-27744646 on Build GRCh38
rs1018915604 Chr.1: 27744783-27744783 on Build GRCh38

View Full Product Details