Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150286724-150286981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIART
Entrez Gene ID
148523
Gene Name
circadian associated repressor of transcription
Gene Aliases
C1orf51, CHRONO, GM129
UniGene
Hs.54680
Gene Chromosome Location
Chr.1: 150282533-150287093 on Build GRCh38
Transcripts
NM_001300838.1 NM_001300839.1 NM_001300840.1 NM_001300841.1 NM_144697.3 XM_017000378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1333784 c.1028C>T p.P343L Chr.1: 150286824-150286824 on Build GRCh38
3934077 c.936C>A p.V312V Chr.1: 150286732-150286732 on Build GRCh38
4513254 c.931C>T p.P311S Chr.1: 150286727-150286727 on Build GRCh38
3473357 c.937C>T p.P313S Chr.1: 150286733-150286733 on Build GRCh38
6120959 c.938C>A p.P313Q Chr.1: 150286734-150286734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782153706 Chr.1: 150286937-150286937 on Build GRCh38
rs782106255 Chr.1: 150286895-150286895 on Build GRCh38
rs781844173 Chr.1: 150286796-150286796 on Build GRCh38
rs782397015 Chr.1: 150286880-150286880 on Build GRCh38
rs782811450 Chr.1: 150286769-150286769 on Build GRCh38
rs781879596 Chr.1: 150286919-150286919 on Build GRCh38
rs782283824 Chr.1: 150286815-150286815 on Build GRCh38
rs781870923 Chr.1: 150286942-150286942 on Build GRCh38
rs377604128 Chr.1: 150286887-150286887 on Build GRCh38
rs782669189 Chr.1: 150286803-150286803 on Build GRCh38
rs781866351 Chr.1: 150286772-150286772 on Build GRCh38
rs996794948 Chr.1: 150286933-150286933 on Build GRCh38
rs200527176 Chr.1: 150286863-150286863 on Build GRCh38
rs782526535 Chr.1: 150286774-150286774 on Build GRCh38
rs782145542 Chr.1: 150286761-150286761 on Build GRCh38
rs782424847 Chr.1: 150286958-150286958 on Build GRCh38
rs782403086 Chr.1: 150286859-150286859 on Build GRCh38
rs369544642 Chr.1: 150286948-150286948 on Build GRCh38
rs782442348 Chr.1: 150286797-150286797 on Build GRCh38
rs782498280 Chr.1: 150286823-150286823 on Build GRCh38
rs374981213 Chr.1: 150286862-150286862 on Build GRCh38
rs781937144 Chr.1: 150286743-150286743 on Build GRCh38
rs139391722 Chr.1: 150286938-150286938 on Build GRCh38
rs782573429 Chr.1: 150286846-150286846 on Build GRCh38
rs782213892 Chr.1: 150286831-150286831 on Build GRCh38
rs372988503 Chr.1: 150286839-150286839 on Build GRCh38
rs201718059 Chr.1: 150286849-150286849 on Build GRCh38
rs782472189 Chr.1: 150286947-150286947 on Build GRCh38
rs782614903 Chr.1: 150286949-150286949 on Build GRCh38
rs782649652 Chr.1: 150286825-150286825 on Build GRCh38
rs782736910 Chr.1: 150286794-150286794 on Build GRCh38
rs369334170 Chr.1: 150286871-150286871 on Build GRCh38
rs782190710 Chr.1: 150286890-150286890 on Build GRCh38
rs1041817374 Chr.1: 150286805-150286805 on Build GRCh38

View Full Product Details