Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81670360-81670585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC137
Entrez Gene ID
339230
Gene Name
coiled-coil domain containing 137
Gene Aliases
RaRF
UniGene
Hs.744125
Gene Chromosome Location
Chr.17: 81666697-81673907 on Build GRCh38
Transcripts
NM_199287.2 XM_011524738.2 XM_017024573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4669750 c.489delA p.A166fs*7 Chr.17: 81670445-81670445 on Build GRCh38
5821556 c.464A>T p.K155M Chr.17: 81670420-81670420 on Build GRCh38
293613 c.488_489insA p.A166fs*9 Chr.17: 81670444-81670445 on Build GRCh38
3523666 c.497+1G>A p.? Chr.17: 81670454-81670454 on Build GRCh38
3664808 c.445G>A p.E149K Chr.17: 81670401-81670401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762660080 Chr.17: 81670384-81670384 on Build GRCh38
rs751322931 Chr.17: 81670393-81670393 on Build GRCh38
rs909966835 Chr.17: 81670528-81670528 on Build GRCh38
rs778335489 Chr.17: 81670466-81670466 on Build GRCh38
rs74808058 Chr.17: 81670427-81670427 on Build GRCh38
rs544918829 Chr.17: 81670530-81670530 on Build GRCh38
rs542449820 Chr.17: 81670539-81670539 on Build GRCh38
rs747511134 Chr.17: 81670470-81670470 on Build GRCh38
rs74002500 Chr.17: 81670518-81670518 on Build GRCh38
rs961391007 Chr.17: 81670479-81670479 on Build GRCh38
rs7215601 Chr.17: 81670535-81670535 on Build GRCh38
rs933674149 Chr.17: 81670507-81670507 on Build GRCh38
rs769412116 Chr.17: 81670382-81670382 on Build GRCh38
rs768317709 Chr.17: 81670494-81670494 on Build GRCh38
rs779033426 Chr.17: 81670453-81670453 on Build GRCh38
rs1001726229 Chr.17: 81670397-81670397 on Build GRCh38
rs750781558 Chr.17: 81670420-81670420 on Build GRCh38
rs548789959 Chr.17: 81670546-81670546 on Build GRCh38
rs957815363 Chr.17: 81670436-81670436 on Build GRCh38
rs771603702 Chr.17: 81670471-81670471 on Build GRCh38
rs55814747 Chr.17: 81670522-81670522 on Build GRCh38
rs763706159 Chr.17: 81670392-81670392 on Build GRCh38
rs1054666684 Chr.17: 81670527-81670527 on Build GRCh38
rs761547990 Chr.17: 81670416-81670416 on Build GRCh38
rs781667051 Chr.17: 81670560-81670560 on Build GRCh38
rs1010706639 Chr.17: 81670452-81670452 on Build GRCh38
rs972498377 Chr.17: 81670485-81670485 on Build GRCh38
rs775115136 Chr.17: 81670474-81670474 on Build GRCh38
rs753960750 Chr.17: 81670424-81670424 on Build GRCh38
rs763418221 Chr.17: 81670423-81670423 on Build GRCh38
rs891643897 Chr.17: 81670534-81670534 on Build GRCh38
rs748831871 Chr.17: 81670484-81670484 on Build GRCh38
rs1039813008 Chr.17: 81670538-81670538 on Build GRCh38
rs758703145 Chr.17: 81670465-81670465 on Build GRCh38
rs911170455 Chr.17: 81670521-81670521 on Build GRCh38
rs766996668 Chr.17: 81670481-81670484 on Build GRCh38
rs375302711 Chr.17: 81670383-81670383 on Build GRCh38
rs773883168 Chr.17: 81670495-81670495 on Build GRCh38
rs767426074 Chr.17: 81670417-81670417 on Build GRCh38
rs573143099 Chr.17: 81670419-81670419 on Build GRCh38
rs752280937 Chr.17: 81670504-81670504 on Build GRCh38
rs201831605 Chr.17: 81670454-81670454 on Build GRCh38

View Full Product Details