Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 76346335-76346606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS10
Entrez Gene ID
79738
Gene Name
Bardet-Biedl syndrome 10
Gene Aliases
C12orf58
UniGene
Hs.96322
Gene Chromosome Location
Chr.12: 76344486-76348442 on Build GRCh38
Transcripts
NM_024685.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3465529 c.1513C>T p.P505S Chr.12: 76346472-76346472 on Build GRCh38
3465530 c.1461A>G p.L487L Chr.12: 76346524-76346524 on Build GRCh38
1184499 c.1628A>G p.N543S Chr.12: 76346357-76346357 on Build GRCh38
1240888 c.1508A>G p.Y503C Chr.12: 76346477-76346477 on Build GRCh38
5425993 c.1617A>G p.P539P Chr.12: 76346368-76346368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142297424 Chr.12: 76346390-76346390 on Build GRCh38
rs200718870 Chr.12: 76346573-76346573 on Build GRCh38
rs759185809 Chr.12: 76346537-76346538 on Build GRCh38
rs572088920 Chr.12: 76346420-76346420 on Build GRCh38
rs1048483339 Chr.12: 76346491-76346491 on Build GRCh38
rs77565309 Chr.12: 76346440-76346440 on Build GRCh38
rs751664217 Chr.12: 76346478-76346478 on Build GRCh38
rs966837486 Chr.12: 76346534-76346534 on Build GRCh38
rs778694077 Chr.12: 76346518-76346518 on Build GRCh38
rs762271852 Chr.12: 76346458-76346458 on Build GRCh38
rs150467742 Chr.12: 76346368-76346368 on Build GRCh38
rs746529935 Chr.12: 76346547-76346547 on Build GRCh38
rs763861552 Chr.12: 76346445-76346445 on Build GRCh38
rs201962535 Chr.12: 76346422-76346422 on Build GRCh38
rs755869502 Chr.12: 76346406-76346406 on Build GRCh38
rs780123311 Chr.12: 76346392-76346392 on Build GRCh38
rs765785183 Chr.12: 76346378-76346378 on Build GRCh38
rs557474406 Chr.12: 76346462-76346462 on Build GRCh38
rs773864735 Chr.12: 76346452-76346452 on Build GRCh38
rs763473990 Chr.12: 76346567-76346567 on Build GRCh38
rs777077443 Chr.12: 76346448-76346448 on Build GRCh38
rs901748236 Chr.12: 76346417-76346417 on Build GRCh38
rs779567118 Chr.12: 76346543-76346543 on Build GRCh38
rs898539189 Chr.12: 76346490-76346490 on Build GRCh38
rs751574303 Chr.12: 76346576-76346576 on Build GRCh38
rs754529978 Chr.12: 76346481-76346481 on Build GRCh38
rs886049850 Chr.12: 76346513-76346513 on Build GRCh38
rs939024065 Chr.12: 76346524-76346524 on Build GRCh38
rs770556842 Chr.12: 76346383-76346386 on Build GRCh38
rs1021050605 Chr.12: 76346522-76346522 on Build GRCh38
rs745860265 Chr.12: 76346519-76346519 on Build GRCh38
rs786204705 Chr.12: 76346533-76346537 on Build GRCh38
rs760364553 Chr.12: 76346444-76346444 on Build GRCh38
rs577177579 Chr.12: 76346463-76346463 on Build GRCh38
rs770239827 Chr.12: 76346558-76346558 on Build GRCh38
rs774417303 Chr.12: 76346431-76346431 on Build GRCh38
rs869025211 Chr.12: 76346438-76346438 on Build GRCh38
rs138434761 Chr.12: 76346549-76346549 on Build GRCh38
rs761098896 Chr.12: 76346363-76346363 on Build GRCh38
rs543722869 Chr.12: 76346508-76346508 on Build GRCh38
rs138215518 Chr.12: 76346559-76346559 on Build GRCh38
rs750402316 Chr.12: 76346383-76346383 on Build GRCh38
rs757789222 Chr.12: 76346374-76346374 on Build GRCh38
rs56867176 Chr.12: 76346471-76346471 on Build GRCh38
rs745357628 Chr.12: 76346432-76346432 on Build GRCh38
rs373327878 Chr.12: 76346426-76346426 on Build GRCh38
rs750994616 Chr.12: 76346469-76346469 on Build GRCh38
rs771936058 Chr.12: 76346521-76346521 on Build GRCh38
rs201701959 Chr.12: 76346548-76346548 on Build GRCh38
rs762251582 Chr.12: 76346553-76346553 on Build GRCh38
rs753629989 Chr.12: 76346495-76346495 on Build GRCh38
rs961208138 Chr.12: 76346507-76346507 on Build GRCh38
rs146812823 Chr.12: 76346395-76346395 on Build GRCh38
rs35676114 Chr.12: 76346369-76346369 on Build GRCh38
rs771662934 Chr.12: 76346437-76346437 on Build GRCh38
rs886043841 Chr.12: 76346474-76346475 on Build GRCh38
rs201022680 Chr.12: 76346479-76346479 on Build GRCh38
rs886049849 Chr.12: 76346418-76346418 on Build GRCh38
rs1057517031 Chr.12: 76346443-76346443 on Build GRCh38

View Full Product Details