Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41316439-41316700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC97
Entrez Gene ID
90324
Gene Name
coiled-coil domain containing 97
Gene Aliases
-
UniGene
Hs.437497
Gene Chromosome Location
Chr.19: 41309855-41324883 on Build GRCh38
Transcripts
NM_052848.1 XM_005259377.4 XM_017027442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078494 c.334C>T p.R112C Chr.19: 41316671-41316671 on Build GRCh38
1200111 c.331T>C p.F111L Chr.19: 41316668-41316668 on Build GRCh38
4078493 c.303C>T p.H101H Chr.19: 41316640-41316640 on Build GRCh38
997007 c.335G>A p.R112H Chr.19: 41316672-41316672 on Build GRCh38
4078492 c.295C>T p.L99L Chr.19: 41316632-41316632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746631058 Chr.19: 41316490-41316490 on Build GRCh38
rs756558548 Chr.19: 41316461-41316461 on Build GRCh38
rs1054550975 Chr.19: 41316658-41316658 on Build GRCh38
rs143841115 Chr.19: 41316524-41316524 on Build GRCh38
rs776221674 Chr.19: 41316503-41316503 on Build GRCh38
rs779195307 Chr.19: 41316483-41316483 on Build GRCh38
rs772622272 Chr.19: 41316584-41316584 on Build GRCh38
rs765298122 Chr.19: 41316500-41316500 on Build GRCh38
rs762910887 Chr.19: 41316613-41316613 on Build GRCh38
rs75624130 Chr.19: 41316665-41316665 on Build GRCh38
rs142783394 Chr.19: 41316671-41316671 on Build GRCh38
rs373386903 Chr.19: 41316550-41316550 on Build GRCh38
rs148083057 Chr.19: 41316551-41316551 on Build GRCh38
rs138157924 Chr.19: 41316552-41316552 on Build GRCh38
rs139158128 Chr.19: 41316640-41316640 on Build GRCh38
rs533020104 Chr.19: 41316600-41316600 on Build GRCh38
rs1010484646 Chr.19: 41316482-41316482 on Build GRCh38
rs771296615 Chr.19: 41316479-41316479 on Build GRCh38
rs781344568 Chr.19: 41316595-41316595 on Build GRCh38
rs1024726317 Chr.19: 41316518-41316518 on Build GRCh38
rs747840097 Chr.19: 41316593-41316593 on Build GRCh38
rs759242580 Chr.19: 41316626-41316626 on Build GRCh38
rs201081145 Chr.19: 41316683-41316683 on Build GRCh38
rs763043456 Chr.19: 41316538-41316538 on Build GRCh38
rs749828863 Chr.19: 41316477-41316477 on Build GRCh38
rs376802126 Chr.19: 41316650-41316650 on Build GRCh38
rs757734565 Chr.19: 41316557-41316557 on Build GRCh38
rs755865258 Chr.19: 41316681-41316681 on Build GRCh38
rs761392998 Chr.19: 41316505-41316505 on Build GRCh38
rs541346433 Chr.19: 41316649-41316649 on Build GRCh38
rs770783004 Chr.19: 41316621-41316621 on Build GRCh38
rs768197651 Chr.19: 41316501-41316501 on Build GRCh38
rs1007369681 Chr.19: 41316523-41316523 on Build GRCh38
rs199582362 Chr.19: 41316666-41316666 on Build GRCh38
rs746229440 Chr.19: 41316575-41316575 on Build GRCh38
rs778241651 Chr.19: 41316462-41316462 on Build GRCh38
rs769492052 Chr.19: 41316506-41316506 on Build GRCh38
rs377096486 Chr.19: 41316597-41316597 on Build GRCh38
rs751897410 Chr.19: 41316652-41316652 on Build GRCh38
rs777870937 Chr.19: 41316553-41316553 on Build GRCh38
rs760937643 Chr.19: 41316641-41316641 on Build GRCh38
rs763469952 Chr.19: 41316615-41316632 on Build GRCh38
rs200453165 Chr.19: 41316625-41316625 on Build GRCh38
rs780984453 Chr.19: 41316592-41316592 on Build GRCh38
rs368463774 Chr.19: 41316634-41316634 on Build GRCh38
rs766152235 Chr.19: 41316533-41316533 on Build GRCh38
rs754348188 Chr.19: 41316554-41316554 on Build GRCh38
rs150772304 Chr.19: 41316676-41316676 on Build GRCh38
rs201218332 Chr.19: 41316542-41316542 on Build GRCh38
rs377395901 Chr.19: 41316541-41316541 on Build GRCh38
rs147264663 Chr.19: 41316558-41316558 on Build GRCh38
rs751004878 Chr.19: 41316675-41316675 on Build GRCh38
rs149006709 Chr.19: 41316511-41316511 on Build GRCh38
rs368669358 Chr.19: 41316674-41316674 on Build GRCh38
rs775446181 Chr.19: 41316555-41316555 on Build GRCh38
rs764617716 Chr.19: 41316639-41316649 on Build GRCh38
rs866465919 Chr.19: 41316615-41316615 on Build GRCh38

View Full Product Details