Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40303086-40303311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A2
Entrez Gene ID
1298
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
UniGene
Hs.418012
Gene Chromosome Location
Chr.1: 40300487-40317653 on Build GRCh38
Transcripts
NM_001852.3 XM_006710365.3 XM_011540715.2 XM_011540716.2 XM_011540717.2 XM_017000332.1 XM_017000333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6575316 c.1549-3C>T p.? Chr.1: 40303188-40303188 on Build GRCh38
4753489 c.1564G>C p.D522H Chr.1: 40303170-40303170 on Build GRCh38
1342617 c.1553G>A p.R518Q Chr.1: 40303181-40303181 on Build GRCh38
4007919 c.1549-1G>C p.? Chr.1: 40303186-40303186 on Build GRCh38
2169114 c.1586C>T p.A529V Chr.1: 40303148-40303148 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762429171 Chr.1: 40303167-40303167 on Build GRCh38
rs752181838 Chr.1: 40303116-40303116 on Build GRCh38
rs769009288 Chr.1: 40303165-40303165 on Build GRCh38
rs371602612 Chr.1: 40303219-40303219 on Build GRCh38
rs755341417 Chr.1: 40303236-40303236 on Build GRCh38
rs200949058 Chr.1: 40303181-40303181 on Build GRCh38
rs901123053 Chr.1: 40303179-40303179 on Build GRCh38
rs770628561 Chr.1: 40303147-40303147 on Build GRCh38
rs755572801 Chr.1: 40303120-40303120 on Build GRCh38
rs146113977 Chr.1: 40303144-40303144 on Build GRCh38
rs371463096 Chr.1: 40303115-40303115 on Build GRCh38
rs768412299 Chr.1: 40303113-40303113 on Build GRCh38
rs142974512 Chr.1: 40303135-40303135 on Build GRCh38
rs752590752 Chr.1: 40303216-40303216 on Build GRCh38
rs201661697 Chr.1: 40303173-40303173 on Build GRCh38
rs972066637 Chr.1: 40303283-40303283 on Build GRCh38
rs1024554840 Chr.1: 40303276-40303276 on Build GRCh38
rs150020975 Chr.1: 40303170-40303170 on Build GRCh38
rs1013130976 Chr.1: 40303263-40303263 on Build GRCh38
rs757702675 Chr.1: 40303235-40303235 on Build GRCh38
rs767322883 Chr.1: 40303193-40303193 on Build GRCh38
rs140697524 Chr.1: 40303183-40303183 on Build GRCh38
rs753444465 Chr.1: 40303226-40303226 on Build GRCh38
rs991450935 Chr.1: 40303252-40303252 on Build GRCh38
rs189183321 Chr.1: 40303250-40303250 on Build GRCh38
rs754855540 Chr.1: 40303104-40303104 on Build GRCh38
rs763536147 Chr.1: 40303223-40303223 on Build GRCh38
rs756853823 Chr.1: 40303229-40303229 on Build GRCh38
rs909958775 Chr.1: 40303249-40303249 on Build GRCh38
rs34110948 Chr.1: 40303199-40303199 on Build GRCh38
rs778665729 Chr.1: 40303232-40303232 on Build GRCh38
rs761700412 Chr.1: 40303278-40303278 on Build GRCh38
rs958842601 Chr.1: 40303259-40303259 on Build GRCh38
rs780267285 Chr.1: 40303150-40303150 on Build GRCh38
rs777296729 Chr.1: 40303124-40303124 on Build GRCh38
rs760533824 Chr.1: 40303105-40303106 on Build GRCh38
rs141556170 Chr.1: 40303158-40303158 on Build GRCh38
rs773987348 Chr.1: 40303182-40303182 on Build GRCh38

View Full Product Details