Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37237808-37238053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFAF7
Entrez Gene ID
55471
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 7
Gene Aliases
C2orf56, MidA, PRO1853
UniGene
Hs.433466
Gene Chromosome Location
Chr.2: 37231631-37259052 on Build GRCh38
Transcripts
NM_001083946.1 NM_144736.4 XM_005264410.1 XM_005264411.1 XM_011532956.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667622 c.371G>A p.G124D Chr.2: 37237830-37237830 on Build GRCh38
1020728 c.349G>A p.A117T Chr.2: 37237808-37237808 on Build GRCh38
6091645 c.379A>G p.R127G Chr.2: 37237838-37237838 on Build GRCh38
4544783 c.363G>A p.V121V Chr.2: 37237822-37237822 on Build GRCh38
4915936 c.383G>A p.G128E Chr.2: 37237842-37237842 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534383592 Chr.2: 37238023-37238023 on Build GRCh38
rs751024038 Chr.2: 37237835-37237835 on Build GRCh38
rs1048557158 Chr.2: 37237915-37237915 on Build GRCh38
rs748193705 Chr.2: 37237874-37237874 on Build GRCh38
rs376115486 Chr.2: 37237871-37237871 on Build GRCh38
rs765851936 Chr.2: 37237830-37237830 on Build GRCh38
rs112166222 Chr.2: 37237964-37237964 on Build GRCh38
rs910083872 Chr.2: 37237920-37237920 on Build GRCh38
rs775517484 Chr.2: 37237907-37237907 on Build GRCh38
rs115423532 Chr.2: 37237912-37237912 on Build GRCh38
rs749265055 Chr.2: 37237895-37237895 on Build GRCh38
rs554174156 Chr.2: 37237906-37237906 on Build GRCh38
rs775341529 Chr.2: 37237961-37237961 on Build GRCh38
rs36027292 Chr.2: 37238020-37238023 on Build GRCh38
rs200563103 Chr.2: 37237855-37237855 on Build GRCh38
rs144797574 Chr.2: 37237883-37237883 on Build GRCh38
rs887005211 Chr.2: 37238003-37238003 on Build GRCh38
rs376451811 Chr.2: 37237857-37237857 on Build GRCh38
rs751947102 Chr.2: 37237854-37237854 on Build GRCh38
rs75490807 Chr.2: 37237994-37237994 on Build GRCh38
rs938391514 Chr.2: 37237837-37237837 on Build GRCh38
rs556547531 Chr.2: 37237846-37237846 on Build GRCh38
rs201488931 Chr.2: 37238021-37238024 on Build GRCh38
rs149775309 Chr.2: 37238015-37238018 on Build GRCh38
rs770835664 Chr.2: 37237901-37237901 on Build GRCh38
rs891680800 Chr.2: 37238009-37238009 on Build GRCh38
rs11900451 Chr.2: 37237980-37237980 on Build GRCh38
rs780603959 Chr.2: 37237849-37237849 on Build GRCh38
rs368016715 Chr.2: 37237889-37237889 on Build GRCh38
rs777828139 Chr.2: 37237887-37237887 on Build GRCh38
rs745637300 Chr.2: 37237917-37237917 on Build GRCh38
rs998168157 Chr.2: 37237983-37237983 on Build GRCh38

View Full Product Details