Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51456265-51456506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
UniGene
Hs.511386
Gene Chromosome Location
Chr.15: 51447724-51622833 on Build GRCh38
Transcripts
NM_001174116.1 NM_001174117.1 NM_015263.3 XM_005254255.1 XM_005254256.1 XM_017022034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7273715 c.8325T>A p.H2775Q Chr.15: 51456316-51456316 on Build GRCh38
4823661 c.8327A>G p.Q2776R Chr.15: 51456314-51456314 on Build GRCh38
4942842 c.8330A>G p.Y2777C Chr.15: 51456311-51456311 on Build GRCh38
4824878 c.8326C>G p.Q2776E Chr.15: 51456315-51456315 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747678752 Chr.15: 51456409-51456409 on Build GRCh38
rs148767441 Chr.15: 51456428-51456428 on Build GRCh38
rs3736484 Chr.15: 51456386-51456386 on Build GRCh38
rs781053158 Chr.15: 51456342-51456342 on Build GRCh38
rs1013612704 Chr.15: 51456398-51456398 on Build GRCh38
rs772785249 Chr.15: 51456415-51456415 on Build GRCh38
rs76659258 Chr.15: 51456367-51456367 on Build GRCh38
rs887983607 Chr.15: 51456365-51456365 on Build GRCh38
rs764791362 Chr.15: 51456377-51456378 on Build GRCh38
rs753979216 Chr.15: 51456366-51456366 on Build GRCh38
rs777263201 Chr.15: 51456396-51456396 on Build GRCh38
rs769297770 Chr.15: 51456393-51456393 on Build GRCh38
rs3736485 Chr.15: 51456413-51456413 on Build GRCh38
rs184258693 Chr.15: 51456419-51456419 on Build GRCh38
rs754937594 Chr.15: 51456321-51456321 on Build GRCh38
rs553944220 Chr.15: 51456452-51456452 on Build GRCh38
rs888810675 Chr.15: 51456394-51456394 on Build GRCh38
rs750075147 Chr.15: 51456391-51456395 on Build GRCh38
rs780598054 Chr.15: 51456378-51456378 on Build GRCh38
rs989991419 Chr.15: 51456473-51456473 on Build GRCh38
rs751303807 Chr.15: 51456300-51456300 on Build GRCh38
rs758501362 Chr.15: 51456369-51456369 on Build GRCh38
rs141293464 Chr.15: 51456357-51456357 on Build GRCh38
rs79082738 Chr.15: 51456463-51456463 on Build GRCh38
rs780067092 Chr.15: 51456351-51456351 on Build GRCh38
rs1034594894 Chr.15: 51456433-51456433 on Build GRCh38
rs902945054 Chr.15: 51456302-51456302 on Build GRCh38

View Full Product Details