Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74636206-74636447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENC1
Entrez Gene ID
8507
Gene Name
ectodermal-neural cortex 1
Gene Aliases
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10
UniGene
Hs.104925
Gene Chromosome Location
Chr.5: 74627406-74641424 on Build GRCh38
Transcripts
NM_001256574.1 NM_001256575.1 NM_001256576.1 NM_003633.3 NR_046318.1 XM_011543696.2 XM_011543697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
739247 c.123G>A p.Q41Q Chr.5: 74636363-74636363 on Build GRCh38
1644568 c.71A>T p.H24L Chr.5: 74636415-74636415 on Build GRCh38
6653992 c.46G>A p.G16S Chr.5: 74636440-74636440 on Build GRCh38
5734381 c.209_211delACT p.Y70delY Chr.5: 74636275-74636277 on Build GRCh38
1311306 c.70C>T p.H24Y Chr.5: 74636416-74636416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144785597 Chr.5: 74636319-74636319 on Build GRCh38
rs780799806 Chr.5: 74636251-74636251 on Build GRCh38
rs777268285 Chr.5: 74636241-74636241 on Build GRCh38
rs767895962 Chr.5: 74636309-74636309 on Build GRCh38
rs138968666 Chr.5: 74636232-74636232 on Build GRCh38
rs758208474 Chr.5: 74636240-74636240 on Build GRCh38
rs142095580 Chr.5: 74636264-74636264 on Build GRCh38
rs750399028 Chr.5: 74636402-74636402 on Build GRCh38
rs142002005 Chr.5: 74636374-74636374 on Build GRCh38
rs770180987 Chr.5: 74636270-74636270 on Build GRCh38
rs774597799 Chr.5: 74636296-74636296 on Build GRCh38
rs751117981 Chr.5: 74636358-74636358 on Build GRCh38
rs775660875 Chr.5: 74636280-74636280 on Build GRCh38
rs746480154 Chr.5: 74636242-74636242 on Build GRCh38
rs35704961 Chr.5: 74636312-74636313 on Build GRCh38
rs758360992 Chr.5: 74636344-74636344 on Build GRCh38
rs375711383 Chr.5: 74636393-74636393 on Build GRCh38
rs201825413 Chr.5: 74636261-74636261 on Build GRCh38
rs148178379 Chr.5: 74636330-74636330 on Build GRCh38
rs752366801 Chr.5: 74636336-74636336 on Build GRCh38
rs370749999 Chr.5: 74636359-74636359 on Build GRCh38
rs749807753 Chr.5: 74636281-74636281 on Build GRCh38
rs898145670 Chr.5: 74636243-74636243 on Build GRCh38
rs761967252 Chr.5: 74636299-74636299 on Build GRCh38
rs769248815 Chr.5: 74636294-74636294 on Build GRCh38
rs960370545 Chr.5: 74636398-74636398 on Build GRCh38
rs892523386 Chr.5: 74636305-74636305 on Build GRCh38
rs756924157 Chr.5: 74636244-74636244 on Build GRCh38
rs201691105 Chr.5: 74636345-74636345 on Build GRCh38

View Full Product Details