Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60159003-60159270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA4
Entrez Gene ID
762
Gene Name
carbonic anhydrase 4
Gene Aliases
CAIV, Car4, RP17
UniGene
Hs.89485
Gene Chromosome Location
Chr.17: 60149941-60178118 on Build GRCh38
Transcripts
XM_005257639.2 XM_011525183.2 XM_017025012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1172445 c.780delG p.E261fs*4 Chr.17: 60159265-60159265 on Build GRCh38
5741080 c.769T>C p.Y257H Chr.17: 60159254-60159254 on Build GRCh38
5605591 c.756C>T p.F252F Chr.17: 60159241-60159241 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766089463 Chr.17: 60159204-60159204 on Build GRCh38
rs550394772 Chr.17: 60159168-60159168 on Build GRCh38
rs549334435 Chr.17: 60159225-60159225 on Build GRCh38
rs768900579 Chr.17: 60159201-60159201 on Build GRCh38
rs973565442 Chr.17: 60159162-60159162 on Build GRCh38
rs150432787 Chr.17: 60159246-60159246 on Build GRCh38
rs779300156 Chr.17: 60159123-60159123 on Build GRCh38
rs1030289775 Chr.17: 60159075-60159075 on Build GRCh38
rs529983471 Chr.17: 60159193-60159193 on Build GRCh38
rs546924276 Chr.17: 60159195-60159195 on Build GRCh38
rs142401267 Chr.17: 60159072-60159072 on Build GRCh38
rs146362767 Chr.17: 60159202-60159202 on Build GRCh38
rs774043643 Chr.17: 60159205-60159205 on Build GRCh38
rs571708928 Chr.17: 60159118-60159118 on Build GRCh38
rs528142989 Chr.17: 60159042-60159042 on Build GRCh38
rs140515828 Chr.17: 60159238-60159238 on Build GRCh38
rs565056769 Chr.17: 60159112-60159112 on Build GRCh38
rs1021190457 Chr.17: 60159137-60159137 on Build GRCh38
rs566759180 Chr.17: 60159198-60159198 on Build GRCh38
rs1032238872 Chr.17: 60159175-60159175 on Build GRCh38
rs757505800 Chr.17: 60159243-60159243 on Build GRCh38
rs767546771 Chr.17: 60159221-60159221 on Build GRCh38
rs962477037 Chr.17: 60159134-60159134 on Build GRCh38
rs879449973 Chr.17: 60159100-60159100 on Build GRCh38
rs567212140 Chr.17: 60159174-60159174 on Build GRCh38
rs745912445 Chr.17: 60159184-60159184 on Build GRCh38
rs1014094305 Chr.17: 60159052-60159052 on Build GRCh38
rs900775628 Chr.17: 60159050-60159050 on Build GRCh38
rs745887096 Chr.17: 60159145-60159145 on Build GRCh38
rs771018420 Chr.17: 60159024-60159024 on Build GRCh38
rs371711521 Chr.17: 60159037-60159037 on Build GRCh38
rs759093480 Chr.17: 60159216-60159216 on Build GRCh38
rs558803561 Chr.17: 60159226-60159226 on Build GRCh38
rs1003897421 Chr.17: 60159088-60159088 on Build GRCh38
rs569363525 Chr.17: 60159233-60159233 on Build GRCh38
rs966599918 Chr.17: 60159167-60159167 on Build GRCh38

View Full Product Details