Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69167783-69168056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB1
Entrez Gene ID
891
Gene Name
cyclin B1
Gene Aliases
CCNB
UniGene
Hs.23960
Gene Chromosome Location
Chr.5: 69167010-69178245 on Build GRCh38
Transcripts
NM_031966.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3828348 c.22-1G>A p.? Chr.5: 69167907-69167907 on Build GRCh38
1739185 c.36T>G p.N12K Chr.5: 69167922-69167922 on Build GRCh38
7222461 c.105C>A p.S35S Chr.5: 69167991-69167991 on Build GRCh38
1200144 c.131C>T p.A44V Chr.5: 69168017-69168017 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752795547 Chr.5: 69167863-69167863 on Build GRCh38
rs759860075 Chr.5: 69167953-69167953 on Build GRCh38
rs777039051 Chr.5: 69167950-69167950 on Build GRCh38
rs758600094 Chr.5: 69167869-69167869 on Build GRCh38
rs1013381457 Chr.5: 69167868-69167868 on Build GRCh38
rs544873273 Chr.5: 69167829-69167829 on Build GRCh38
rs762054869 Chr.5: 69167827-69167827 on Build GRCh38
rs776092366 Chr.5: 69167957-69167957 on Build GRCh38
rs1034366112 Chr.5: 69168021-69168021 on Build GRCh38
rs560413531 Chr.5: 69168032-69168032 on Build GRCh38
rs748035379 Chr.5: 69168002-69168002 on Build GRCh38
rs1002946303 Chr.5: 69167997-69167997 on Build GRCh38
rs933588052 Chr.5: 69167969-69167969 on Build GRCh38
rs767702393 Chr.5: 69167973-69167973 on Build GRCh38
rs865916737 Chr.5: 69167854-69167854 on Build GRCh38
rs373480903 Chr.5: 69167857-69167857 on Build GRCh38
rs758336269 Chr.5: 69168007-69168007 on Build GRCh38
rs749098045 Chr.5: 69167890-69167890 on Build GRCh38
rs746606130 Chr.5: 69167929-69167929 on Build GRCh38
rs778022575 Chr.5: 69167875-69167875 on Build GRCh38
rs530270969 Chr.5: 69167852-69167852 on Build GRCh38
rs140927235 Chr.5: 69167919-69167919 on Build GRCh38
rs759192061 Chr.5: 69167858-69167858 on Build GRCh38
rs754814106 Chr.5: 69168000-69168000 on Build GRCh38
rs373876739 Chr.5: 69167881-69167881 on Build GRCh38
rs765963344 Chr.5: 69167979-69167979 on Build GRCh38
rs369506933 Chr.5: 69167966-69167966 on Build GRCh38
rs770501273 Chr.5: 69167939-69167939 on Build GRCh38
rs772739694 Chr.5: 69167922-69167922 on Build GRCh38
rs879617924 Chr.5: 69167826-69167828 on Build GRCh38
rs577627491 Chr.5: 69167826-69167826 on Build GRCh38
rs764207072 Chr.5: 69167962-69167962 on Build GRCh38
rs775282499 Chr.5: 69167898-69167904 on Build GRCh38
rs747630808 Chr.5: 69167917-69167917 on Build GRCh38
rs971913060 Chr.5: 69167987-69167987 on Build GRCh38
rs1000750363 Chr.5: 69167878-69167878 on Build GRCh38
rs750148681 Chr.5: 69167956-69167956 on Build GRCh38
rs377039637 Chr.5: 69167910-69167910 on Build GRCh38
rs757429226 Chr.5: 69167889-69167889 on Build GRCh38
rs899494711 Chr.5: 69167871-69167871 on Build GRCh38
rs768630510 Chr.5: 69167905-69167905 on Build GRCh38
rs865987149 Chr.5: 69167983-69167983 on Build GRCh38
rs542396320 Chr.5: 69167867-69167867 on Build GRCh38
rs777205019 Chr.5: 69168014-69168014 on Build GRCh38
rs762338775 Chr.5: 69167963-69167963 on Build GRCh38
rs146181241 Chr.5: 69167851-69167851 on Build GRCh38

View Full Product Details